MilliporeSigma
US EPA Method 8141: GC Analysis of Organophosphorus Pesticides (OP-Pest) on SLB®-5ms
US EPA Method 8141: GC Analysis of Organophosphorus Pesticides (OP-Pest) on SLB®-5ms suitable for GC
Materials
analytical column
Product No.
Description
Pricing
SLB®-5ms Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 0.25 μm
CONDITIONS
column
SLB-5ms, 30 m x 0.25 mm I.D., 0.25 μm (28471-U)
inj. temp.
250 °C
detector
NPD, 320 °C
carrier gas
helium, 1.5 mL/min
injection
2.0 μL, splitless (0.75 min)
liner
4 mm I.D., single taper
sample
nitrogen-phosphorous pesticide standard, each analyte at 100 ppb in MTBE
Description
Legal Information
SLB is a registered trademark of Merck KGaA, Darmstadt, Germany