US EPA Method 8141: GC Analysis of Organophosphorus Pesticides (OP-Pest) on SPB®-608
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
SPB-608, 30 m x 0.25 mm I.D., 0.25 μm (24103-U)
oven
60 °C (1 min), 5 °C/min to 200 °C, 15 °C/min to 300 °C (5 min)
inj. temp.
250 °C
detector
NPD, 320 °C
carrier gas
helium, 1.5 mL/min
injection
2.0 μL, splitless (0.75 min)
liner
4 mm I.D., single taper
sample
nitrogen-phosphorous pesticide standard, each analyte at 100 ppb in MTBE
Description
Legal Information
SPB is a registered trademark of Merck KGaA, Darmstadt, Germany