MilliporeSigma
GC Analysis of Gasoline Residue in Arson Samples on a 100% Methyl Column after SPME using 100 μm PDMS Fiber
GC Analysis of Gasoline Residue in Arson Samples on a 100% Methyl Column after SPME using 100 μm PDMS Fiber suitable for GC, application for SPME
Materials
Related product
Product No.
Description
Pricing
Equity-1 Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 0.25 μm
used together
Product No.
Description
Pricing
SPME Fiber Assembly
100 μm PDMS, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3, red hub
CONDITIONS
SPME fiber
polydimethylsiloxane phase fiber, 100 μm headspace sampling (20 min) 10 sec desorption (splitless mode) (57300-U)
column
100% methyl polysiloxane, 30 m × 0.25 mm I.D., 0.25 μm (Supelco equivalent, Equity-1, 28046-U)
oven
35 °C (2 min) to 220 °C at 10 °C/min, hold 2 min, to 300 °C at 30 °C/min, hold 5 min
inj. temp.
splitless (closed 3 min), 220 °C (2 mm I.D. injector liner)
detector
FID, 300 °C
carrier gas
helium, 1 mL/min (split 50:1)