MilliporeSigma
HPLC Analysis of Aflatoxins in Peanut Paste on Ascentis® Express C18 following SPE using Supel Tox AflaZea
HPLC Analysis of Aflatoxins in Peanut Paste on Ascentis® Express C18 following SPE using Supel™ Tox AflaZea application for HPLC, application for SPE
Materials
accessory
Product No.
Description
Pricing
Micro-Reaction Vessel, black phenolic hole cap, pkg of 12
volume 5 mL, clear glass vial, PTFE/red rubber septum, O.D. × H 20 mm × 62 mm, thread for 20-400
Visiprep SPE Vacuum Manifold
DL (Disposable Liner), 12-port model
related product
Product No.
Description
Pricing
Aflatoxin Mix 4 solution
analytical standard
Ascentis® Express C18 (2.7 μm) HPLC Column
L × I.D. 15 cm × 2.1 mm, pk of 1 ea
CONDITIONS
sample preparation
Solid Phase Extraction
sample/matrix
raw peanut paste
SPE tube/cartridge
Supel Tox AflaZea, 6 mL (55314-U)
extraction process
suspend 25 g raw peanut paste in 100 mL acetonitrile:water (84:16) extraction solution
sample addition
directly load 2 mL extract onto SPE tube
elution
pull sample through the SPE tube by vacuum at a rate of 8 mL/min
eluate post-treatment
combine 200 μL eluate and 300 μL of water:trifluoroacetic acid:acetic acid (70:20:10 ) derivatization reagent, (heat at 65°C for 25 min, add 500 μL of water to derivatized sample and vortex 30 sec)
column
Ascentis Express C18, 15 cm x 2.1 mm I.D., 2.7 μm particles (53825-U)
mobile phase
[A} acetonitrile; {B} water; (20:80, A:B)
flow rate
0.2 mL/min
column temp.
35 °C
detector
fluorescence, ex 365 nm, em 450 nm
injection
20 μL
sample
B1, G1 (8 ppb), B2, G2 (2 ppb)
Description
Analysis Note
Aflatoxins are carcinogenic metabolites produced by fungi that infect major agricultural commodites. The presence and concentrations of aflatoxins is widely regulated. Here, a cleanup method using interference removal SPE followed by analysis by HPLC with fluorescence detection is used to analyze derivatized samples extracted from raw peanut paste. The HPLC method was optimized for fluorescence detection and quantification of aflatoxins at < 10 ppb concentrations.
Other Notes
Sample pretreatment: suspend 25 g raw peanut paste in 100 mL acetonitrile:water (84:16) extraction solution
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany
Supel is a trademark of Sigma-Aldrich Co. LLC