MilliporeSigma
HPLC Analysis of Terfenadine Enantiomers on Astec® Cellulose DMP
HPLC Analysis of Terfenadine Enantiomers on Astec® Cellulose DMP application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Astec® Cellulose DMP Chiral HPLC Column
5 μm particle size, L × I.D. 15 cm × 4.6 mm
mobile phase component
Product No.
Description
Pricing
Diethylamine
≥99.5%
standard
Product No.
Description
Pricing
Terfenadine
CONDITIONS
column
Astec Cellulose DMP, 15 cm x 4.6 mm I.D., 5 μm particles (51098AST)
mobile phase
[A] heptane; [B] ethanol; [C] diethylamine; (98:2:0.1, A:B:C)
flow rate
0.5 mL/min
column temp.
40 °C
detector
UV, 230 nm
injection
3 μL
sample
1 g/L in methanol