LC/MS Analysis of Antiarrhythmics on Ascentis® Express Biphenyl
CONDITIONS
column
Ascentis Express Biphenyl, 10 cm x 3.0 mm, 2.7 μm particles (64066-U)
mobile phase
10 mM ammonium formate (70:30 water:acetonitrile) SW pH 7.0 (not adjusted)
flow rate
0.6 mL/min
pressure
1363 psi (94 bar)
column temp.
35 °C
detector
MS ESI(+), TOF, XIC
injection
5 μL
sample
300 ng/mL in 85:15, water:methanol
Description
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany