MilliporeSigma
UHPLC Analysis of Barbiturates on Titan C18
UHPLC Analysis of Barbiturates on Titan C18 application for UHPLC
Materials
standard
Product No.
Description
Pricing
Uracil
≥99.0%
Barbital
Butalbital solution
1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Phenobarbital solution
1 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
CONDITIONS
column
Titan C18, 10 cm x 2.1 mm I.D., 1.9 μm particles (577124-U)
mobile phase
[A] 0.1% ammonium acetate (not adjusted); [B] acetonitrile; (70:30, A:B)
flow rate
0.4 mL/min
pressure
7540 psi (520 bar)
column temp.
35 °C
detector
UV, 230 nm
Description
Analysis Note
Analysis of a barbiturate mix using an MS-compatible mobile phase and a Titan C18 UHPLC column is shown here. Ultra high purity LC-MS solvents were used to supply low background interference and low particulate contaminants for robust, trouble-free operation. Cerilliant CRMs provided reliable identification and quantification.
Legal Information
null