Merck
PCR in as Little as 15 Minutes!
KOD One™ PCR Master Mix for
Ultra-Fast, High Fidelity PCR
KOD One™ PCR Master Mix
KOD One™ PCR Master Mix is the right choice for ultra-fast PCR. It offers the fastest elongation rates available for high fidelity polymerases without compromising on the yield and specificity. This convenient, premixed, ready-to-use solution offers an elongation rate of up to 5 seconds per kb and exhibits approximately 80-fold higher fidelity than Taq DNA polymerase. Additionally, this flexible master mix will easily fit right into your PCR workflow with its ability to amplify directly from crude samples and use primers containing both uracil and inosine. Choose KOD One™ PCR Master Mix and efficiently amplify your samples with confidence.
Benefits
The KOD OnePCR Master Mix formulation offers the following advanced features:
  • Ready-to-Use
  • Added Flexibility
  • High Fidelity
  • High Efficiency
  • Crude Sample PCR
  • High Specificity
  • Primers with inosine (dI) and uracil (dU)
Applications
This product has a variety of applications such as:
  • DNA/cDNA amplification
  • Direct PCR
  • Colony PCR
  • NGS library amplification
  • PCR amplification to detect site-directed mutagenesis
PCR Master Mix and Polymerase Cross Reference Guide 
ProductQ5® Hot Start High-Fidelity 2X Master MixPhire Hot Start II PCR Master MixPlatinum™ SuperFi PCR Master MixVELOCITY DNA PolymeraseKOD One™ PCR Master Mix
Catalog NumberM0494S/M0494L (100/500 rxns)F125S/F125L (220/1000 rxns)12351010/12351050 (100/500 rxns)BIO-21098/BIO-21099 (125/250 reactions)KMM-101NV/KMM-201NV
Hot start formulationAptamer basedAffibody  inactivation methodAntibody inactivation methodNot hot-start enzymeAntibody inactivation method
Target SizeUp to 20 kbUp to 7.5 kbUp to 20 kbUp to 30 kbUp to 40 kb
Fidelity vs Taq~280X higher than Taq2X higher than Taq>300X higher than Taq50X higher than Taq80X higher than Taq
Elongation rate20-30 sec/kb (complex template) 10 sec/kb (simple template)10 to 15 sec/kb15 to 30 sec/kb1 5sec/kb for ≤5 kb 30 sec/kb for targets up to 10 kb1 sec for <1 kb 5 sec/kb for <10 kb 10 sec/kb for ~10 kb
Related Products
Loading
KOD One™ PCR Master Mix Customer Testimonials
Aleksandar Vještica
Assistant Professor
Organization: University of Lausanne
Field: Academia / University
"Thanks to its high processivity, KOD One™ Master Mix allowed us to quickly clone plasmids using only overlapping PCRs. This approach did not require restriction enzyme digestions of cloning inserts and thus considerably simplified the cloning protocol and saved us time. "
Review date: 3 December 2021
KOD One™ PCR Master Mix
Blanco Lago Solis
Ph.D. Student
Organization: University of Geneva
Field: Academia / University
"I successfully amplified a 14kb fragment in 2h15 vs 4h30 with Phusion or Q5."
KOD One™ PCR Master Mix
Maria Cristina Gambetta
Assistant Professor
Organization: University of Lausanne
Field: Academia / University
"I was caught red-handed in my impatience. I asked around and indeed got an aliquot of KOD One™ master mix promising to give it back. It worked wonderfully! I guess this really is a miraculous polymerase. So, we went ahead and ordered master mix to not waste time, as experiments are urgent."
Review date: 16 December 2021
KOD One™ PCR Master Mix
Dr. Michael Taschner
Senior Scientist
Organization: University of Lausanne
Field: Academia / University
"I received a free sample of KOD One™ PCR master mix to test. I was amazed by the speed, processivity, and accuracy of PCRs carried out on complicated templates. No other product I used in the past was able to amplify a product of >12 kb in record time from ‘quick-and-dirty’ yeast genomic DNA (purified according to [Lōoke et al, BioTechniques, 2011]. I fully recommend this product for difficult PCR cases."
Review date: 17 December 2021
KOD One™ PCR Master Mix