Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

HA00D-100G

Monobind Inc.

Monobind Inc. Human Serum Albumin Lyophilized

Standard Reagent Grade - 100 g

Synonym(s):

HSA, HSA Dried

Sign Into View Organizational & Contract Pricing


About This Item

PDF Icon
Shipped & Sold By Monobind Inc.
A Trusted Marketplace Seller
PDF Icon
Have a question?

form

dry

biological source

human

storage temp.

< 8 °C

storage condition

OK to freeze

manufacturer/tradename

Monobind Inc.

General description

Human Serum Albumin Dried for use in preparing controls, calibrators and standards in various professional RUO-applications such as assay kits, lab analyses, research studies, and clinical trials.

  • Shelf Life: 5 years from DOM at 8°C or Colder.
  • Packaging & Shipping: 100g in Media Bottle. Ships Frozen with Cool Packs, Overnight Only.
  • Purity: >96%
  • Certificates of Analysis (COA): Ensures product has met all performance specifications prior to release for sale and accompanies every lot.
  • Origin & Testing*: US Collection Centers - Donor Testing negative for (HIV-1&2, HCV, HBsAg, HIV-Ag, ALT and RPR) by FDA Method. *May vary by lot, refer to the provided COA.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library