Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

VTSNTR-99

Valutek

Valutek Spunlace Non-Woven Wiper

Polyester Cleanroom Wipe, Cold Knife Cut Edge, 9 in. x 9 in.

Synonym(s):

Polyester wipe, Ployester-Cellulose wipers

Sign Into View Organizational & Contract Pricing


About This Item

PDF Icon
Shipped & Sold By Valutek
A Trusted Marketplace Seller
PDF Icon
Have a question?

manufacturer/tradename

Valutek

General description

VTSNTR is Valutek's spunlace non-woven wiper, is made from a hydro-entangled nonwoven blend consisting of 55% cellulose and 45% polyester.

This composite substrate brings together the excellent absorbent characteristics of a natural fiber with the cleanliness and strength of a synthetic yarn, resulting in a material with minimal levels of particulate and extractable counts.

Every Valutek wipers undergo laundering and packaging within a cleanroom environment. Furthermore, all Valutek wipers are thoroughly tested and produced in ISO-compliant facilities, subject to Valutek's inspection and rigorous process control measures to uphold Valutek's quality standards and product specifications.

Product Features:

  • Comprised of a 55% cellulose and 45% polyester blend.
  • Features a cold knife cut edge and does not contain any chemical binders.
  • Offers high absorbency but has low abrasion resistance.
  • Compatible with common cleaning and disinfecting solutions.
  • Exhibits low levels of extractables and moderate particle counts.
  • Available in a range of sizes to accommodate various applications.
  • Critical environment compatible.
  • Part number and lot number traceable for 36 months from the manufacturing date.

Product Application:

Valutek classifies this product as "Macrotek" in terms of cleanliness level, and recommends it to use for  cleanroom Class 10,000+ (ISO 7+) critical environment. It is specifically engineered for critical processing applications where the highest level of contamination control is essential.

The spunlace non-woven wiper is often referred to as the "multi-tasking" or "grunt" wipe due to its versatility.

It serves as a highly absorbent wiper suitable for general-purpose cleaning in environmental and facility settings. However, it is not recommended for direct contact with products.

Product Packaging:

  • 9" x 9" (300 Ea/Bag) 
  • 12" x 12" (150 Ea/Bag) 
  • 18" x 18" (75 Ea/Bag) 
  • Each wiper is carefully packaged in double poly bags, vacuum-sealed, flat-packed inside carton boxes, and includes a carton liner for added protection.
  • Priced and sold by the bags.

**This wiper substrate with its specific edge treatment is also available in a pre-wetted form.**


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library