MilliporeSigma
HomeServicesSoftware & Digital PlatformsDigital Reaction Optimization with Catalexis
Digital Reaction Optimization with Catalexis
Smarter Chemistry, Faster Results
What if catalyst and ligand screening for complex cross-coupling reactions could be predictable, fast, and AI-driven instead of relying on traditional trial and error workflows? Catalexis delivers exactly that by combining AI driven catalyst optimization, BayBE Bayesian modeling, and a curated 23 ligand phosphine plate to enable smarter, more efficient screening for transformations such as Buchwald-Hartwig aminations and Suzuki–Miyaura couplings.
Built for both traditional benchtop chemistry and high-throughput experimentation (HTE), Catalexis enables researchers to explore reaction space digitally, reduce experimental burden, and accelerate the identification of high performing ligands in cross-coupling reactions, and other sensitive transformations where small parameter shifts impact yields and selectivity.
How does Catalexis work?
Step 1 - Order Your Catalexis Kit
Step 1: Order Your Catalexis Kit
Choose the format that suits your workflow:
50 mg Ligand Kit - 23 phosphine ligands + Pd source + 5 portal tokens.   
1 µmol Screening Plates - five plates (23 ligands each) + Pd source + 5 portal tokens.
Step 2 - Run Your 23‑Ligand Screening Reactions
Run your 23‑ligand screen using your chosen substrate and conditions, keeping all variables constant. Compatible with manual setups and automated HTE workflows.
Step 3 - Upload Your Reaction Results to the Catalexis Portal
Upload your reaction yields (0 -100) into the Catalexis portal and redeem 1 token to receive your top ligand candidates. No chemistry metadata needed.
Step 4 - Generate AI Driven Ligand Rankings Driven Ligand Rankings
Catalexis compares your data against its database of 400+ ligands and highlights top performers. 
Step 5 - Optimize With Guided Recommendations
Define your reaction parameters and let the BayBE (Bayesian back-end) ML engine propose  the most promising next experiments within your chemical space, thereby, allowing you to sample the chemical space more efficiently than traditional DOE approaches.
Step 6 - Explore the Reaction Space with BayBE
Upload your new results and instantly receive suggested next conditions to continue improving yield
Step 7 - Repeat to Refine Results
Iterate steps 5-6 until your reaction achieves the desired target, be it yield, conversion, byproduct limitation or other custom variable.
Why Choose Catalexis Over Traditional Catalyst Screening?
Loading
User Testimonials
Discover how Catalexis is transforming labs – here’s what chemists are saying about their experience:
“Catalexis significantly streamlined our di-amination reaction optimization of 1,3-dichlorobenzene, saving time by providing a curated set of ligands to screen. After three rounds of screening, we identified the best ligand for our final industrial-scale process.”
Pharma Biotech Process Development Lab
“The suggested ligands delivered excellent results, including some I hadn’t initially considered trying.”
Academic Medicinal Chemistry Lab
"Catalexis is a nice tool wherein a kit of 23 ligands has the potential to evaluate and predict the performance of around 500 ligands with the help of AI."
Synthetic R&D Chemist
Related Resources
Request Form
To contact us and learn more about the platform, please fill out the form with your dedicated request. Fields marked with an asterisk (*) are mandatory.