HPLC Analysis of Heroin, Cocaine, Amphetamine and MDMA on Ascentis® Express HILIC
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
Ascentis Express HILIC, 10 cm x 2.1 mm I.D., 2.7 μm particles (53939-U)
mobile phase
[A] 10 mM ammonium acetate in water, pH 3.5; [B] 10 mM ammonium acetate in acetonitrile; (2.98, A:B)
flow rate
0.4 mL/min
column temp.
50 °C
detector
ESI(+), m/z 100-400
injection
1 μL
Description
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany