Merck
HomeqPCRSYBR® Green Extract-N-Amp™ PCR Kit
SYBR® Green Extract-N-Amp™ PCR Kit
Product Description
The SYBR® Green Extract-N-Amp™ Tissue PCR Kit contains all the reagents needed for rapid extraction, amplification and detection of genomic DNA from mouse tails and other animal tissues, buccal swabs, hair shafts, and saliva. DNA is rapidly extracted from a tissue by incubating the sample with a mixture of the Extraction Solution and the Tissue Preparation Solution at room temperature for 10 minutes. There is no need for freezing tissue in liquid nitrogen, mechanical disruption, organic extraction, column purification or precipitation of the DNA. After a 3-minute heat denaturing step, an equal volume of Neutralization Solution B is added to the extract to neutralize inhibitory substances and the extract is ready for real-time PCR in any plate-based real-time thermal cycling system.
An aliquot of the neutralized extract is then combined with the Extract-N-Amp™ SYBR® Green PCR ReadyMix™ and user-provided PCR primers. The Extract-N-Amp™ SYBR® Green PCR ReadyMix is a 2X reaction mix containing SYBR® Green, buffer, salts, dNTPs, Taq polymerase and JumpStart™ Taq antibody. It is optimized specifically for use with the extraction reagents and contains JumpStart Taq antibody for hot start PCR to enhance specificity and SYBR® Green I to act as a nonspecific reporter for realtime PCR.
Reagents ProvidedProduct No.XNATG
100 extractions
and 100
amplifications
XNATRG
1,000 extractions
and 1,000
amplifications
Extraction SolutionE752624 mL240 mL
Neutralization Solution BN391024 mL240 mL
Tissue Preparation SolutionT30733.0 mL30 mL
Extract-N-Amp SYBR Green PCR
ReadyMix. This is a 2X real-time PCR
reaction mix
 containing SYBR Green,
buffer, salts, dNTPs, Taq polymerase
and JumpStart Taq antibody.
S43201.2 mL12 mL
Reagents and Equipment Required but Not Provided
  • Microcentrifuge tubes (1.5 or 2 mL) or multiwell plate for extractions
  • Small dissecting scissors or scalpel
  • Forceps (small to medium in size)
  • Buccal swab (Sterile foam tipped applicator, Product No. A9601)
  • Tubes or plate for PCR
  • Heat block or thermal cycler at 95 °C
  • PCR Primers
  • Plate-based Thermal cycler capable of real-time SYBR® Green detection.
  • Reference dye, if required for real-time thermal cycler
  • Water, PCR grade, Product No. W1754
Precautions and Disclaimer
This product is for R&D use only, not for drug, household, or other uses. Please consult the Safety Data Sheet for information regarding hazards and safe handling practices.
Storage of Kit Components
The Extraction, Tissue Preparation & Neutralization Solutions can be stored at 2-8 °C up to 3 weeks; if storing longer than 3 weeks, keep the solutions at -20 °C. Do not store in a "frost-free" freezer.
The Extract-N-Amp™ SYBR® Green PCR ReadyMix should always be kept at –20 °C and out of light as much as possible. Excessive freeze-thawing of the ReadyMix can be detrimental to product performance. Aliquot the ReadyMix into suitably sized portions if necessary to avoid more than five freeze-thaws.
Preliminary Considerations
Primer Design & Optimization
SYBR® Green I will detect both specific and non-specific PCR amplicons. Well-designed primers are recommended to ensure the highest possible specificity. Primers for PCR should be designed with the aid of primer design software to eliminate the complications introduced with primer-dimers, nonspecific hybridization and secondary structures. The size of the PCR target should preferably be less than 500 bp, although this product has performed with targets up to 1000 bp. Larger targets are often harder to quantify via real-time PCR.
The primer concentration and cycling parameters need to be optimized and will depend on the system being used. Typical final primer concentrations are ~0.4 µM each. Lower primer concentrations may decrease the accumulation of primer-dimer formation and nonspecific product formation.
For further information on primer design and optimization, consult the manufacturer of your thermalcycling system for assistance.
Controls
A positive control is always helpful to verify that the PCR reaction is performing properly. Use purified genomic DNA from the same species as the extract and dilute to 1-5 ng/μL with a 50:50 mixture of Extraction and Neutralization Solutions or Extract-N-Amp™ PCR Diluent (Product No. E8155). Do not use water to dilute the positive control. Replace the 4 μL of extract with 4 μL of the positive control in a 20 μL PCR reaction. See Section C., Step 1.
A negative control is necessary to determine if contamination or primer-dimer formation is present. Replace the 4 μL of extract with 4 μL of a 50:50 mixture of Extraction and Neutralization Solutions or Extract-N-Amp™ PCR Diluent in a 20 μL PCR reaction. See Section C., Step 1. Do not use water as a negative control.
Procedure
All steps are carried out at room temperature unless otherwise noted. The Extraction, Neutralization and Tissue Preparation solutions should be completely thawed, brought to room temperature and gently mixed before use. The Extract-N-Amp™ SYBR® Green PCR ReadyMix should be thawed, mixed well, but kept on ice until needed.
A. DNA extraction from Mouse Tails, Animal Tissues, Hair, or Saliva
  1. Pipette 100 µL of Extraction Solution into a microcentrifuge tube or well of a multiwell plate. Add 25 µL of Tissue Preparation Solution to the tube or well and pipette up and down to mix.
  2. Note:
  3. 2a. For Fresh or Frozen Mouse Tails: Rinse the scissors or scalpel and forceps in 70% ethanol prior to use and between different samples. Place a 0.5-1 cm piece of mouse tail tip, cut end down, into the Extraction/Tissue Preparation Solution mixture. Ensure that the cut end of the mouse tail is completely submerged in the solution.
    Note: Frozen tails may be cut and used as they thaw. For fresh mouse tails, perform extractions within 30 minutes of snipping the tail.
    2b. For Animal tissues: Rinse the scissors or scalpel and forceps in 70% ethanol prior to use and between different samples. Place a 2 – 10 mg piece of tissue into the Extraction/Tissue Preparation Solution mixture. Ensure that the tissue is completely submerged in the solution.
    2c. For Hair Shafts: Rinse the scissors and forceps in 70% ethanol prior to use and between different samples. Trim excess off of the hair shaft, leaving the root and place sample (root end down) into the Extraction/Tissue Preparation Solution mixture. Only one hair shaft, with root, is required per extraction.
    2d. For Saliva: Pipette 10 µL of saliva into the Extraction/Tissue Preparation Solution mixture. Mix thoroughly by vortexing or pipetting.
  4. Incubate sample at room temperature for 10 minutes.
  5. Incubate sample at 95 °C for 3 minutes. Tissues will not be completely digested at the end of the incubations. This is normal and will not affect performance.
    Note: This is a critical step! To ensure the best possible extraction use a heat block or water bath that fits the contour of the tube used for extraction. Heating in an oven does not provide efficient heat conduction. Increase incubation time if a heat block or water bath is not available. Additional time needed must be determined empirically.
  6. Add 100 µL of Neutralization Solution B to heated extract and mix by vortexing.
  7. Store the neutralized tissue extract at 4 °C ,or use immediately in PCR – Section C, Step 1.
    Note: For long term storage, remove the undigested tissue or transfer the extracts to new tubes or wells. Extracts may now be stored at 4 °C for at least 6 months without notable loss in most cases.
B. DNA extraction for Buccal Swabs
  1. Collect buccal cells on swab and allow the swab to dry. Drying time is approximately 10 to 15 minutes.
    Note: Due to the low volume of solution used for DNA extraction, a foam tipped swab should be used. Swabs with fibrous tips, such as cotton or dacron, should be avoided because the solution can not be recovered efficiently.
  2. Pipette 200 µL of Extraction Solution into a 1.5 mL microcentrifuge tube. Add 25 µL of Tissue Preparation Solution to the tube and pipette up and down to mix.
    Note: If several extractions will be performed, sufficient volumes of Extraction and Tissue Preparation Solutions may be pre-mixed in a ratio of 8:1 up to 2 hours before use.
  3. Place dried buccal swab into the solution and incubate at room temperature for 1 minute.
  4. Twirl swab in solution 10 times and then remove excess solution from the swab into the tube by twirling swab firmLy against the side of the tube. Discard the swab. Close the tube and vortex briefly.
  5. Incubate sample at room temperature for 10 minutes.
  6. Incubate sample at 95 °C for 3 minutes.
    Note: This is a critical step! To ensure the best possible extraction use a heat block or water bath that fits the contour of the tube used for extraction. Heating in an oven does not provide efficient heat conduction. Increase incubation time if a heat block or water bath is not available. Additional time needed must be determined empirically.
  7. Add 200 µL of Neutralization Solution B to heated extract and mix by vortexing.
  8. Store the neutralized extract at 4 °C, or use immediately in PCR – Section C, Step 1.
    Note: Extracts may be stored at 4 °C for at least 6 months without notable loss in most cases.
C. Real-Time PCR Amplification
The Extract-N-Amp™ SYBR® Green PCR ReadyMix contains JumpStart Taq antibody for specific hot start amplification. Therefore, PCR reactions can be assembled at room temperature without premature Taq DNA polymerase activity. See reference 1 for general information regarding real-time PCR.
With some tissues or other starting materials, PCR may be inhibited by secondary metabolites in the extract such that little or no detectable PCR product is obtained. Diluting the extract five-, ten- or twenty-fold with a 50:50 mixture of Extraction and Neutralization Solutions or with Extract-N-Amp™ PCR Diluent has been shown to alleviate PCR inhibition in many cases.
Note: The reagents included in this kit were formulated together and are dependent on one another for optimal performance of this product. Therefore, only use the recommended diluents instead of water to dilute extracts.
  1. Prepare Reaction Mixture
    Add the following reagents to a thin-walled PCR microcentrifuge tube or plate appropriate for the instrument to be used. Be aware that many instruments require an internal reference dye, which should be included in the reaction mixture.
ReagentVolume / 1 RXN
Extract-N-Amp SYBR Green PCR ReadyMix10 µL
Water, PCR Gradex µL
Forward Primery µL
Reverse Primery µL
Reference dye*z µL*
Neutralized Extract4 µL
Total Volume20 µL
PCR Reaction Setup
* If required for real-time PCR instrument.
Alternatively, the total volume of a PCR reaction can be adjusted by scaling the reagents relative to the desired PCR total volume.
When preparing multiple PCR reactions, it may be beneficial to create a master-mix. Generate a volume in excess of the amount required to account for measuring losses. After determining the appropriate volume for all reagents, combine and gently vortex to create master-mix. Aliquot 16 µL of master mix and 4 µL of extract per reaction.
    2.    Mix & Spin Down
           Mix gently and briefly centrifuge to collect all components at the bottom of the tube or plate.
    3.    Amplification Parameters
           The amplification parameters should be optimized for individual primers, template, and thermal cyclers
           with real-time SYBR® Green detection.
StepTemperatureTimeCycles
Initial
Denaturation
94 °C3 minutes1
Denaturation94 °C15 seconds
– 1 minute
30-40
Annealing45 to 68 °C15 seconds
– 1 minute
Extension72 °C10 seconds
– 1 minute
Common Cycling Parameters
4.   Data Analysis
          Follow the recommendations of the real-time instrument used to effectively analyze results.
          Run a melt curve to ensure the correct PCR target was amplified1. For further verification, run the PCR
          reaction on a 2% Agarose-TBE buffered gel.
Troubleshooting Guide
ProblemPossible CauseSolution
No PCR product (no fluorescence detected) in sample and positive controlThe primers may not be designed optimally.Confirm the accuracy of the sequence information. If the primers are less than 22 nucleotides long, try to lengthen the primer to 25-30 nucleotides. If the primer has a GC content of less than 45%, try to redesign the primer with a GC content of 45-60%.
A PCR component may be missing or degradedA checklist is recommended when assembling reactions. Ensure the DNA isn’t degraded in the positive control. Run a 2% Agarose TBEbuffered gel to confirm if the target was amplified, but not detected.
The annealing temperature may be too high.Decrease the annealing temperature in 2-4 °C increments.
The extension time may be too short.
Increase the extension time in 30 second increments, especially for longer templates.
The denaturation temperature may be too high or too low.
Optimize the denaturation temperature by increasing or decreasing the temperature in 1 °C increments.
The denaturation time may be too long or too short.Optimize the denaturation time by increasing or decreasing the time in 10 second increments.
There may be too few cycles performed.Increase the number of cycles (5-10 additional cycles at a time).
Target template is difficult.
In most cases, inherently difficult targets are due to unusually high GC content and/or secondary structure. Betaine has been reported to help amplification of high GC content templates at a concentration of 1.0-1.7 M.
No PCR product (signal) in sample, but there is product
in positive control
The DNA extraction
was inadequate
Repeat the extraction; thaw all kit reagents, bring to room temperature and thoroughly mix all kit reagents prior to use. Increase length of incubation and/or temperature if normal extraction protocol is inadequate.
Heating in step 4 was inadequate
Verify that extracts were heated in a water bath or a block that fits the contour of tubes, and that the bath or block was at 95 °C for the entire 3 minute incubation. Use of other heating devices, such as an oven, will require longer incubation times that must be determined empirically.
Extract was stored without removing tail snip or tissue.
Remove undigested tail snip or tissue before storing extracts at 4 °C. Extracted DNA may not be stable in the presence of undigested tissue.
PCR reaction may be inhibited due to secondary metabolites in the extract.
Dilute the extract 5-, 10-, or 20-fold with a 50:50 mix of Extraction and Neutralization Solutions or Extract-N-Amp PCR Diluent (Product Code E 8155) and repeat PCR with 4 µl of diluted extract.
Multiple PCR products
JumpStart Taq antibody is not working correctly.
Do not use DMSO or formamide with Extract-N-Amp SYBR Green PCR ReadyMix. It can interfere with the enzyme-antibody complex. Other cosolvents, solutes (e.g., salts) and extremes in pH or other reaction conditions may reduce the affinity of the JumpStart Taq antibody for Taq polymerase and thereby compromise its effectiveness.
Multiple PCR products (continued)
Touchdown PCR may be needed.“Touchdown” PCR significantly improves the specificity of many PCR reactions in various applications. Touchdown PCR uses an annealing/extension temperature that is higher than the Tm of the primers during the initial PCR cycles. The annealing/extension temperature is then reduced to the primer Tm for the remaining PCR cycles. The change can be performed in a single step or in increments over several cycles. Note that fluorescence is inversely proportional to temperature, thus fluorescence must be read at the same temperature every cycle.
Primer-dimers are co-amplified
Include an additional step in the cycling program at a temperature slightly below the Tm of the desired PCR product (approximately 3 °C) where fluorescence is read to avoid detection of primer-dimers.
Primer concentration is too high.Reduce the primer concentration in a series of two-fold dilutions, i.e., 0.2 µM, 0.1 µM, 0.05 µM, and test in a trial set of PCR reactions.
Primers are degraded.Check for primer degradation on a polyacrylamide gel.
Negative Control shows PCR product (signal)Reagents or reactions have been
contaminated
Extraneous DNA template may have been introduced into the reagents or when setting up the PCR reactions. Clean the area in which PCR is setup. Then, rerun the experiment being careful not to contaminate the reactions. If product still amplifies in negative controls, the reagents were probably contaminated and should be replaced with unused
reagents.
Materials
Loading
References
1.
Ririe KM, Rasmussen RP, Wittwer CT. 1997. Product Differentiation by Analysis of DNA Melting Curves during the Polymerase Chain Reaction. Analytical Biochemistry. 245(2):154-160. https://doi.org/10.1006/abio.1996.9916
NOTICE TO PURCHASER: LIMITED LICENSE
Use of this product is covered by one or more of the following US patents and corresponding patent claims outside  the US: 5,994,056 and 6,171,785. The purchase of this product includes a limited, non-transferable immunity from suit under the foregoing patent claims for using only this amount of product for the purchaser’s own internal  research. No right under any other patent claim (such as apparatus or system claims in US Patent No. 6,814,934) and no right to perform commercial services of any kind, including without limitation reporting the results of purchaser's activities for a fee or other commercial consideration, is conveyed expressly, by implication, or by estoppel. This product is for research use only. Diagnostic uses under Roche patents require a separate license from Roche. Further information on purchasing licenses may be obtained by contacting the Director of Licensing, Applied Biosystems, 850 Lincoln Centre Drive, Foster City, California 94404, USA.
SYBR® Green I and its use protected under U.S. Patent No. 5,338,671 and 5,587,287 and corresponding patents in other countries.
SYBR is a registered trademark of Life Technologies
Extract-N-Amp, JumpStart and ReadyMix are trademarks of Sigma-Aldrich Co. LLC
JC, RC,PHC 01/13-1