MilliporeSigma
HomeEnzyme Activity AssaysEnzymatic Assay of α-Amylase (EC 3.2.1.1)
Enzymatic Assay of α-Amylase (EC 3.2.1.1)
Determination of Alpha-Amylase Activity
This procedure may be used for the determination of α-Amylase activity.
The spectrophotometric stop reaction determination (A540, Light path = 1 cm) is based on the following reaction:
Alpha-Amylase
Unit Definition: One unit will liberate 1.0 mg of maltose from starch in 3 minutes at pH 6.9 at 20 °C.
Reagents and Equipment Required
  • Sodium phosphate, monobasic, Product No. S0751
  • Sodium chloride, Product No. S9888
  • Starch from potato, Product No. S2004
  • Sodium hydroxide, Product No. S5881
  • Potassium sodium tartrate, tetrahydrate, Product No. S2377
  • 3,5-Dinitrosalicylic acid, Product No. D0550
  • D‑(+)‑Maltose, monohydrate, Product No. M9171
Precautions
Please consult the Safety Data Sheet for information regarding hazards and safe handling practices.
Preparation Instructions
Use ultrapure water (≥18 MΩ×cm resistivity at 25 °C) for the preparation of reagents.
Buffer solution (20 mM Sodium Phosphate with 6.7 mM Sodium Chloride, pH 6.9 at 20 °C) – Prepare a solution containing 2.4 mg/mL of sodium phosphate, monobasic, Product No. S0751, and 0.39 mg/mL of sodium chloride, Product No. S9888, in ultrapure water.  Adjust to pH 6.9 at 20 °C using 1 M NaOH/1 M HCl.
Starch solution [1.0% (w/v) Soluble Starch Solution] – Prepare a 10 mg/mL solution using starch from potato, Product No. S2004, in Buffer:
  • Solubilize the solution by boiling on a heating/stir plate for 15 minutes with mixing.
  • Remove from heat. Continue to mix the solution and allow to cool to room temperature.
  • Bring the solution to final volume with ultrapure water.
  • Continue mixing the solution throughout the assay procedure.
2 M Sodium Hydroxide (NaOH) solution – Prepare a 80 mg/mL solution using sodium hydroxide, Product No. S5881, in ultrapure water.
5.3 M potassium sodium tartrate, tetrahydrate solution – Prepare a 1,496 mg/mL solution of potassium sodium tartrate, tetrahydrate, Product No. S2377, in 2 M Sodium Hydroxide (NaOH) solution.  Dissolve solids by heating on a heating/stir plate with mixing. Do not heat to a boil!
96 mM 3,5-Dinitrosalicylic acid solution – Prepare a 21.9 mg/mL solution using 3,5‑Dinitrosalicylic acid, Product No. D0550, in ultrapure water.  Dissolve solids by heating on a heating/stir plate with mixing. Do not heat to a boil!
Color Reagent solution – For preparation of 40 mL, add:
  • 12.0 mL of warm (50–70 °C) ultrapure water to an appropriate size amber bottle.
    With mixing, slowly add:
  • 8.0 mL of warm 5.3 M potassium sodium tartrate, tetrahydrate solution
  • 20 mL of warm 96 mM 3,5-Dinitrosalicylic acid solution
This solution is stable for 6 months at ambient temperature if protected from light. Volume prepared may be adjusted if needed.
0.2% (w/v) Maltose Standard – Prepare a 2 mg/mL solution in a volumetric flask using D‑(+)‑maltose, monohydrate, Product No. M9171, in ultrapure water.
α-Amylase Sample solution – Immediately before use, prepare a solution containing 0.75‑1.5 units/mL of α-Amylase in 20 °C ultrapure water.
Procedure
Final assay concentration – In a 2.00 mL reaction volume, the final concentration is 0.50% (w/v) starch and ~1 unit of α-amylase.
1. α-Amylase Sample Assay
a. Pipette (in mL) the following reagents into suitable containers:
ReagentSample 1Sample 2Sample 3Sample Blank
Starch solution1.001.001.001.00
b. Mix by swirling and equilibrate to 20 °C.  Then add (in mL):
ReagentSample 1Sample 2Sample 3Sample Blank
α-Amylase Sample0.500.701.00
c. Mix by swirling and incubate for exactly 3.0 minutes at 20 °C.  Then add:
ReagentSample 1Sample 2Sample 3Sample Blank
Color Reagent1.001.001.001.00
d. Cover containers with a vented cap and place in a boiling water bath for exactly 15 minutes.
e. Remove containers from boiling water bath. Then add (in mL):
ReagentSample 1Sample 2Sample 3Sample Blank
α-Amylase Sample0.500.301.00
f. Cool solutions on ice to room temperature.
g. Then add (in mL):
ReagentSample 1Sample 2Sample 3Sample Blank
Ultrapure water9.009.009.009.00
h. Mix by inversion. Blank a suitable spectrophotometer against air at 540 nm and record the A540 for the Samples and Sample Blank.
2. Standard Curve Preparation
a. Prepare a standard curve by pipetting (in mL) the following reagents into suitable containers:
ReagentSTD1STD2STD3STD4STD5STD6STD7STD BLK
0.2% (w/v) Maltose Standard0.050.200.400.600.801.002.00
Ultrapure water1.951.801.601.401.201.002.00
Color Reagent1.001.001.001.001.001.001.001.00
Note: Additional standard levels may be added as needed.
b. Cover containers with a vented cap and place in a boiling water bath for exactly 15 minutes.
c. Remove containers from boiling water bath. Cool solutions on ice to room temperature.
d. Then add (in mL):
ReagentSTD1STD2STD3STD4STD5STD6STD7STD BLK
Ultrapure water9.009.009.009.009.009.009.009.00
e. Mix by inversion. Blank a suitable spectrophotometer against air at 540 nm and record the A540 for the Standards and Standard Blank.
Results
Calculations
  1. Determine the ΔA540 of each Standard vs. the Standard Blank.
    ΔA540 (Standard) = A540 (Standard) – A540 (Standard Blank)
  2. Prepare a standard curve by plotting the ΔA540 of the standards vs. mg of maltose using linear regression.
  3. Determine the ΔA540 of each Sample vs. the Sample Blank.
    ΔA540 (Sample) = A540 (Sample) – A540 (Sample Blank)
  4. Determine the mg of Maltose released using the standard curve.
  5.     
amylase-eq2
where:
df = dilution factor
mL enzyme  = mL of Sample added in step 1b.
6.
amylase-eq3
Related Products
Loading
Reference
1.
Bernfeld P. 1955. [17] Amylases, ? and ?.149-158. https://doi.org/10.1016/0076-6879(55)01021-5