Merck
HomeSmall Molecule HPLCEphedrine/Pseudoephedrine HCl in Oral Solution Analysis
Ephedrine/Pseudoephedrine HCl in Oral Solution Analysis
Dean Duan
MilliporeSigma China Application Lab Shanghai, China
Chinese Pharmacopeia Method
Ephedrine hydrochloride and pseudoephedrine hydrochloride can be used to reduce cold, allergic rhinitis, rhinitis and sinusitis caused by nasal congestion symptoms and control bronchial asthma. The same compounds are also found in the Chinese traditional medicine Xiao’er Kechuanling oral solution.
This application focuses on the determination of ephedrine HCl and pseudoephedrine HCl. Oral solution samples were distilled to extract the ephedrine HCl and pseudoephedrine HCl. Samples were then filtered through Millex® PTFE syringe filters, prior to HPLC-UV analysis using a Purospher® STAR RP-18 endcapped HPLC column.
The limit of detection (LOD) and the limit of quantitation (LOQ) were 0.04 µg/kg and 0.12 µg/kg, respectively, for ephedrine, and 0.02 µg/kg and 0.06 µg/kg, respectively, for pseudoephedrine. The method can be used for determination of ephedrine HCl and pseudoephedrine HCl in Xiao’er Kechuanling oral solution.
2-D chemical structures (bond line structure) of ephedrine HCl and pseudoephedrine HCl compounds used to determine analytes in Xiao’er Kechuanling oral solution
Figure 1.Chemical structures of compounds used in the study.
Materials
Loading
Determination of Ephedrine HCl and Pseudoephedrine HCl in Xiao’er Kechuanling Oral Solution
Purospher® STAR RP-18e
Experimental Conditions
Column Used:Purospher® STAR RP-18e 250 x 4.6 mm, 5 µm (1.51456.0001)
Detection:Waters e2695, 210 nm
Mobile phase A:10 mM KH2PO4, adjust pH value to 2.3 with H3PO4
Mobile phase B:Methanol
Gradient:A : B = 89 : 11, v/v
Flow Rate: 1.0 mL/min
Injection Volume:20 µL
Temp:30 °C
StandardDissolve appropriate amount of ephedrine HCl and pseudoephedrine HCl in mobile phase to obtain a solution containing 5 µg/mL of ephedrine HCl and 3 µg/mL of pseudoephedrine HCl
Sample:Accurately weigh 5 mL of oral solution into a 500-mL round-bottomed flask, add into 120 mL of 10% sodiμm hydroxide, shake well, distill, collect ~95 mL distillate into a 100-mL volμmetric flask containing 5 mL of 10 mM of 10 mM KH2PO4 (adjust pH value to 2.3 with H3PO4). Make up to the tick mark with water. Filter with 0.45 µm filter membrane
Pressure drop:2150 psi
Table 1.HPLC conditions.
Separation of ephedrine HCl and pseudoephedrine HCl standards used to determine analytes in Xiao’er Kechuanling oral solution by HPLC-UV method
Figure 2.Chromatogram of ephedrine HCl and pseudoephedrine HCl standard solution.
Chromatogram showing standard peaks of Xiao’er Kechuanling oral solution used to determine analytes in Xiao’er Kechuanling oral solution by HPLC-UV method
Figure 3.Chromatogram of in Xiao’er Kechuanling oral solution.
Chromatographic run of solvent blank in HPLC-UV separation of ephedrine HCl and pseudoephedrine HCl standards used to determine analytes in Xiao’er Kechuanling oral solution
Figure 4.Chromatogram of solvent blank.
NoCompoundRetention Time (min)ResolutionTheoretical PlatesTailing Factor
1ephedrine HCl18.2--96981.1
2pseudoephedrine HCl20.02.4102921.1
Table 2.Specificity data of ephedrine HCl and pseudoephedrine HCl.
Specificity and Repeatability – Ephedrine and Pseudoephedrine
Specificity: Inject reference solution and determine the retention time of desired analyte in presence of other components like impurities.
NoCompoundRetention Time (min)Area (%)Tailing FactorRRT
1ephedrine HCl18.256.41.11.0
2pseudoephedrine HCl20.032.81.11.1
Table 3.Specificity data of ephedrine and pseudoephedrine HCl.
Standard repeatability (5 µg/mL) - ephedrine HCl
STD 1212571
STD 2227584
STD 3215716
STD 4212961
STD 5251431
Mean1547859
Standard Deviation16480.3
RSD (%)1.1
Table 4.Standard repeatability data of ephedrine HCl.
Standard Repeatability (5 µg/mL) - Pseudoephedrine HCl
STD 1123655
STD 2123017
STD 3126350
STD 4122884
STD 5133801
Mean1327102
Standard Deviation4611.6
RSD (%)0.3
Table 5.Standard repeatability data of pseudoephedrine HCl.
Calibration Data - Ephedrine
Linearity (area mAU*min), ephedrine HCl
Conc. (µg/mL)Mean Area
0.103380
0.208013
0.4016834
0.520141
1.035864
2.079213
4.0166302
5.0212571
6.0272766
Table 6.Linear range of ephedrine HCl.
LOD and LOQ, ephedrine HCl
Conc. (µg/mL)Mean Area (mAU*min)
0.103380
0.208013
0.4016834
0.5020141
STEYEX508.2
Slope42344.5
LOD (ppm)0.04
LOQ (ppm)0.12
Table 7.LOD and LOQ of ephedrine HCl.
Calibration curve of ephedrine HCl used to measure the analyte in Xiao’er Kechuanling oral solution using Purospher® STAR RP-18e colμmn
Figure 5.Calibration curve of ephedrine HCl.
Calibration Data - Pseudoephedrin
Linearity (area mAU*min), pseudoephedrine HCl
Conc. (µg/mL)Mean Area
0.061976
0.124684
0.248952
0.3011653
0.6019605
1.245571
2.498038
3.0123655
4.0179102
Table 8.Linear range of pseudoephedrine HCl.
4.2 LOD and LOQ - pseudoephedrine HCl
Conc. (µg/mL)Mean Area (mAU*min)
0.061976
0.124684
0.248952
0.3011653
STEYEX255.2
Slope39369.9
LOD (ppm)0.02
LOQ (ppm)0.06
Table 9.LOD and LOQ of pseudoephedrin HCl.
Calibration curve of pseudoephedrine HCl to measure the analyte in Xiao’er Kechuanling oral solution using Purospher® STAR RP-18e colμmn
Figure 6.Calibration curve of pseudoephedrine HCl.
Conclusion
The method can be used for ephedrine HCl and pseudoephedrine HCl in Xiao’er Kechuanling oral solution.