MilliporeSigma
HomeCross-CouplingChiral Quest Phosphine Ligands
Chiral Quest Phosphine Ligands
One of the most efficient methods for constructing chiral compounds is asymmetric hydrogenation. Catalytic asymmetric hydrogenations are among the most widely used industrial catalytic processes, due to their high turnover rates, efficiency and atom economy, and inexpensive material costs. Transition metal complexes associated with chiral phosphorus ligands are the dominant choice of catalysts for asymmetric hydrogenation, in large part due to the Nobel prize-winning, pioneering work of Noyori and Knowles. The requirement of an electron-rich chiral phosphine ligand is at the core of this transformation.
Professor Xumu Zhang at Penn State has made remarkable advances by creating a toolbox of chiral phosphines which can be used on a variety of substrates, some of which have been historically resistant to facile hydrogenation. Furthermore, an additional benefit in some reductions is reduced catalyst loading, due to increased turnover numbers (TON). Sigma-Aldrich is pleased to announce an agreement with Chiral Quest to distribute research quantities of a series of Zhang’s chiral phosphines for catalytic asymmetric hydrogenations.
C3-TunePhos, a member of the atropisomeric aryl bisphosphine ligand family with tunable dihedral angles, provides comparable or superior enantioselectivities and catalytic abilities to BINAP in Ru-catalyzed asymmetric hydrogenation of ß-keto esters (Scheme 1),1 cyclic ß-(acylamino) acrylates (Scheme 2),2 and α-phthalimide ketones (Scheme 3).3
A highly electron-donating, low molecular weight, and rigid P-chiral bisphospholane ligand, TangPhos proves highly efficient in the rhodium-catalyzed hydrogenation of a variety of functionalized olefins such as α-dehydroamino acids (Scheme 4),4 α-arylenamides (Scheme 5),4 ß-(acylamino)acrylates (Scheme 6),5 itaconic acids (Scheme 7),6 and enol acetates (Scheme 8).6
BINAPINE, a highly electron-donating rigid ligand, demonstrates excellent enantioselectivity and reactivity, with TON up to 10,000 for the asymmetric hydrogenation of Z-ß-aryl(ß-acylamino) acrylates (Scheme 9).7
(R)-BINAPHANE shows excellent enantioselectivity (up to >99% ee) for hydrogenation of E/Z-isomeric mixtures of ß-substituted arylenamides (Scheme 10).8
Materials
Loading
References
1.
Zhang Z, Qian H, Longmire J, Zhang X. 2000. Synthesis of Chiral Bisphosphines with Tunable Bite Angles and Their Applications in Asymmetric Hydrogenation of ?-Ketoesters. J. Org. Chem.. 65(19):6223-6226. https://doi.org/10.1021/jo000462v
2.
Tang W, Wu S, Zhang X. 2003. Enantioselective Hydrogenation of Tetrasubstituted Olefins of Cyclic ?-(Acylamino)acrylates. J. Am. Chem. Soc.. 125(32):9570-9571. https://doi.org/10.1021/ja035777h
3.
Lei A, Wu S, He M, Zhang X. 2004. Highly Enantioselective Asymmetric Hydrogenation of ?-Phthalimide Ketone:  An Efficient Entry to Enantiomerically Pure Amino Alcohols. J. Am. Chem. Soc.. 126(6):1626-1627. https://doi.org/10.1021/ja039153n
4.
Tang W, Zhang X. 2002. A chiral 1,2-bisphospholane ligand with a novel structural motif: applications in highly enantioselective Rh-catalyzed hydrogenations. Angew. Chem. Int. Ed. Engl.. 411612.
5.
Tang W, Zhang X. 2002. Highly Efficient Synthesis of Chiral ?-Amino Acid Derivatives via Asymmetric Hydrogenation. Org. Lett.. 4(23):4159-4161. https://doi.org/10.1021/ol026935x
6.
Tang W, Liu D, Zhang X. 2003. Asymmetric Hydrogenation of Itaconic Acid and Enol Acetate Derivatives with the Rh-TangPhos Catalyst. Org. Lett.. 5(2):205-207. https://doi.org/10.1021/ol0272592
7.
Tang W, Wang W, Chi Y, Zhang X. 2003. A Bisphosphepine Ligand with Stereogenic Phosphorus Centers for the Practical Synthesis of ?-Aryl-?-Amino Acids by Asymmetric Hydrogenation. Angew. Chem. Int. Ed.. 42(30):3509-3511. https://doi.org/10.1002/anie.200351465
8.
Xiao D, Zhang Z, Zhang X. 1999. Synthesis of a Novel Chiral Binaphthyl Phospholane and Its Application in the Highly Enantioselective Hydrogenation of Enamides. Org. Lett.. 1(10):1679-1681. https://doi.org/10.1021/ol991074m