MilliporeSigma
DIOP for Asymmetric Hydrogenation Catalysis
DIOP (Product No. 237655; [(-)-2,3-O-Isopropylidene-2,3-dihydroxy-1,4-bis(diphenylphosphino)butane]) is a C2 chelating diphosphine ligand for transition metal complexes during asymmetric catalysis and is one of the first chiral ligands used for asymmetric hydrogenation catalysis. DIOP can be synthesized in four simple steps from tartaric acid. Chelation with a rhodium complex generates an optically active catalyst active in hydrogenation. To demonstrate the activity of DIOP, Kagan and Dang conducted the asymmetric hydrogenation of a variety of alkenes.1 Optical yields of up to 72% were obtained with substrate to catalyst ratio of up to 200:1 (Scheme 1).
DIOP
Scheme 1.
Materials
Loading
References
1.
Dang TP, Kagan HB. 1971. The asymmetric synthesis of hydratropic acid and amino-acids by homogeneous catalytic hydrogenation. J. Chem. Soc. D.(10):481. https://doi.org/10.1039/c29710000481