MilliporeSigma
All Photos(3)
Key Documents
MBD6000
Sigma-Aldrich
5R-Plex kit
Ultra-sensitive 16S NGS assay for degraded and low biomass DNA
Sign Into View Organizational & Contract Pricing
About This Item
UNSPSC Code:
41106302
NACRES:
NA.84
Quality Level
usage
Preparing 96 samples for sequencing
technique(s)
DNA amplification: suitable
DNA sequencing: suitable
shipped in
dry ice
storage temp.
-10 to -25°C
General description
The 16S ribosomal RNA gene (16S rRNA) is a common bacterial marker, used as a target for microbial community profiling in microbiome metagenomic studies. The gene is composed of nine variable regions (V1-V9) interspersed between conserved regions. The most common 16S rRNA NGS assays target one or two regions using a single set of primers (e.g., V3-V4). However, this approach usually results in a poor bacterial detection and classification rate when the DNA input is of low-quality or when applying it on extremely low biomass samples.  The 5R-PLEX kit offers a complete solution to challenging biological samples where the standard 16S sequencing protocols fail to perform.   Sample types include:
  • Formalin-Fixed, Paraffin-Embedded (FFPE) tissue
  • Cancerous tumor tissue 
  • Degraded or damaged DNA  
  • Low biomass samples 
  • Fossil-derived and ancient DNA
The 5R-PLEX kit was developed in collaboration with Dr. Ravid Straussman of the Weissmann Institute, based off a technique that was featured in his Science publication focused on the human tumor microbiome characterization (Science2020 May 29;368(6494):973-980. doi: 10.1126/science.aay9189) . The kit targets five short variable regions along the 16S rRNA gene (V2, V3, V5, V6, V8) that are co-amplified in a multiplexed PCR within a single tube.   After sequencing, the output data is uploaded to our M-CAMPTM platform (Microbiome Computational Analysis for Multi-omics Profiling) and analyzed using the unique 5R-PLEX algorithm[2]. This new analysis approach computationally combines the amplified regions to reconstruct one coherent high-resolution microbial profile. Read more about our kit here.
Features and Benefits
  • The 5R-PLEX Kit provides high profile sequencing from degraded, damaged or low biomass samples
  • Multiplexed NGS assay targets 5 variable regions of the 16S rRNA gene in a single primer pool
  • Includes positive control for trouble shooting experiments.
  • The kit can detect as low as 1pg of highly degraded bacterial DNA with an average fragment size of 260bp.
  • Kit includes low bioburden reagents
  • Includes exclusive access to M-CAMP platform featuring an innovative algorithm designed to reconstruct a single coherent microbial profiling and perform comprehensive analysis.
Components
  • 5R-PLEX PCR1 Primer mix- 25 μL
  • 5R-PLEX PCR2 Forward Primer mix- 25 μL
  • 5R-PLEX index plate 96-plate
  • Water, microbial DNA-free 10X- 1.5 mL
  • HF DNA Polymerase- 0.1 mL
  • 5X HF buffer 2 mL
  • dNTP′s- 0.2 mL
  • Elution Buffer (EB), microbial DNA-free- 8 mL
  • 5R-PLEX Positive Control (10 ng/μL)- 30 μL
Storage and Stability
The shelf life of all reagents provided in the kit is 12 months when stored properly. Store all components at -20 °C.
Other Notes
Kit component details can be found in the User guide.
Legal Information
M-Camp is a trademark of Merck KGaA, Darmstadt, Germany
hcodes
Hazard Classifications
Aquatic Chronic 3
Storage Class
10 - Combustible liquids
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Visit the Document Library
Amnon Amir et al.
Nucleic acids research, 41(22), e205-e205 (2013-11-12)
The emergence of massively parallel sequencing technology has revolutionized microbial profiling, allowing the unprecedented comparison of microbial diversity across time and space in a wide range of host-associated and environmental ecosystems. Although the high-throughput nature of such methods enables the
Deborah Nejman et al.
Science (New York, N.Y.), 368(6494), 973-980 (2020-05-30)
Bacteria were first detected in human tumors more than 100 years ago, but the characterization of the tumor microbiome has remained challenging because of its low biomass. We undertook a comprehensive analysis of the tumor microbiome, studying 1526 tumors and
Nicole M Davis et al.
Microbiome, 6(1), 226-226 (2018-12-19)
The accuracy of microbial community surveys based on marker-gene and metagenomic sequencing (MGS) suffers from the presence of contaminants-DNA sequences not truly present in the sample. Contaminants come from various sources, including reagents. Appropriate laboratory practices can reduce contamination, but
Jacob T Nearing et al.
Microbiome, 9(1), 113-113 (2021-05-20)
Advances in DNA sequencing technology have vastly improved the ability of researchers to explore the microbial inhabitants of the human body. Unfortunately, while these studies have uncovered the importance of these microbial communities to our health, they often do not
Garold Fuks et al.
Microbiome, 6(1), 17-17 (2018-01-28)
Most of our knowledge about the remarkable microbial diversity on Earth comes from sequencing the 16S rRNA gene. The use of next-generation sequencing methods has increased sample number and sequencing depth, but the read length of the most widely used
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service