MilliporeSigma
HomeBiosensors & BioimagingGraphene Quantum Dots: Properties, Synthesis & Applications
Graphene Quantum Dots: Properties, Synthesis & Applications
Introduction to Carbon and Graphene Quantum Dots
Colloidal semiconductor quantum dots (QDs) have numerous potential applications in solar cells, light emitting diodes, bioimaging, electronic displays, and other optoelectronic devices due to their unique size-dependent electro-optical properties, and have thus been of significant research interest.
However, due to the high market cost of inorganic QDs, on the order of thousands of US dollars per gram, their industrial use has been slow and limited. In addition, application development has been hindered by the high toxicity of inorganic QDs. As a promising cost-effective alternative, carbon quantum dots (CDs, CQDs or C-dots) and graphene quantum dots (GQDs) have recently emerged as a new class of QD materials. CDs and GQDs have advantages of nontoxicity, good solubility, stable photoluminescence, and better surface grafting, thus making them promising candidates for replacing inorganic QDs. Moreover, the recent discovery of a one-step multigram synthesis of GQDs from coal and other carbon sources opens the possibility of their large-scale industrial production.
Synthesis of Graphene Quantum Dots
Previous methods of GQD synthesis involved high-cost raw materials such as graphene1 or photonic crystals2 and fairly low-yield and expensive methods such as laser ablation,3 electron beam lithography,4 or electrochemical synthesis.5 These factors made GQDs virtually unavailable for commercial applications. More recent research, reports the preparation of GQD from fairly inexpensive organic sources such as citric acid/urea6 that offers product cost reduction and availability on a larger scale. However, the synthesis of GQDs from coal7 (the least expensive material known) increases the possibility of the use of GQDs in future commercial products. Due to their low production cost, coal-derived GQDs are feasible for large-scale industrial applications and might be successfully used as a cost-effective and eco-friendly alternative to conventional inorganic quantum dots.
In a typical patented process, coal is stirred in concentrated nitric acid and heated at 100o-120 oC for few hours. The solution is cooled, and the nitric acid is evaporated and reused. The GQDs are then filtered using cross-flow ultra-filtration. After purification, the solution is concentrated using rotary evaporation to obtain solid GQDs.
Characterization of GQDs
A variety of high quality GQDs can be produced by controlling the manufacturing process parameters such as raw materials, temperature, and reaction time.
Figure 1 shows representative optical and TEM images of blue luminescent GQDs (Product No. 900708). These images show that the GQDs forms a translucent and stable suspension in water, and typically exhibit disk-shaped structures with a diameter of <5 nm with topographic height of 1–2.0 nm.
Representative optical and TEM images of blue luminescent GQDs
Figure 1. Representative optical and TEM images of blue luminescent GQDs. (a) Optical image of 1 Liter of concentrated GQDs suspension. (b) Optical image of diluted GQDs suspension under visible (left) and 365nm UV light (right). (c) Typical TEM image of GQDs. Inset: HR-TEM image of GQD.
Typical photoluminescent (PL) and UV-VIS properties of GQDs (Figure 2) and the PL properties of GQDs offered in our catalog (Table 1).
UV-VIS properties of GQDs
Figure 2. UV-VIS properties of GQDs. (a) Excitation and emission contour map of GQDs. (b) Photoluminescence emission of GQDs excited at 350nm. (c) Absorption spectra of GQDs.
VioletBlue
(900708, 900726)
Cyan
(900707)
Aqua Green
(900712, 900713)
Quantum Yield (%)>70>70>30>35
Maximum excitation (nm)320350420485
Maximum emission (nm)404445490525
Table 1. Photoluminescent properties of GQDs
Applications of GQDs
In contrast to classic QDs, GQDs are biocompatible, photo-stable, with enhanced surface grafting, and inherit superior thermal, electrical, and mechanical properties from graphene. These features can greatly contribute to various state-of-the-art applications including:
  • Taggants for security/anti-counterfeiting/brand protection applications8
  • Bioimaging markers9
  • Fluorescent polymers10
  • Antibacterial,11 Antibiofouling12 and Disinfection systems.13
  • Heavy Metals,14 Humidity and Pressure sensors15
  • Batteries16
  • Flash memory devices17
  • Photovoltaic devices18
  • Light-emitting diodes19
Summary
Due to the limited availability of GQDs, applications involving them are still being developed and to this end the synthesis of GQDs from coal appears promising as it allows the production of high quality material at a larger scale. The availability of high-quality GQDs in larger quantities to the scientific community will help drive more in-depth studies of the unique properties, as well as accelerate the development of new applications.
Materials
Loading
References
1.
Pan D, Zhang J, Li Z, Wu M. 2010. Hydrothermal Route for Cutting Graphene Sheets into Blue-Luminescent Graphene Quantum Dots. Adv. Mater.. 22(6):734-738. https://doi.org/10.1002/adma.200902825
2.
Guo X, Wang C, Yu Z, Chen L, Chen S. 2012. Facile access to versatile fluorescent carbon dots toward light-emitting diodes. Chem. Commun.. 48(21):2692. https://doi.org/10.1039/c2cc17769b
3.
Sun Y, Zhou B, Lin Y, Wang W, Fernando KAS, Pathak P, Meziani MJ, Harruff BA, Wang X, Wang H, et al. 2006. Quantum-Sized Carbon Dots for Bright and Colorful Photoluminescence. J. Am. Chem. Soc.. 128(24):7756-7757. https://doi.org/10.1021/ja062677d
4.
Li L, Wu G, Yang G, Peng J, Zhao J, Zhu J. 2013. Focusing on luminescent graphene quantum dots: current status and future perspectives. Nanoscale. 5(10):4015. https://doi.org/10.1039/c3nr33849e
5.
Li Y, Hu Y, Zhao Y, Shi G, Deng L, Hou Y, Qu L. 2011. An Electrochemical Avenue to Green-Luminescent Graphene Quantum Dots as Potential Electron-Acceptors for Photovoltaics. Adv. Mater.. 23(6):776-780. https://doi.org/10.1002/adma.201003819
6.
Li X, Zhang S, Kulinich SA, Liu Y, Zeng H. 2015. Engineering surface states of carbon dots to achieve controllable luminescence for solid-luminescent composites and sensitive Be2+ detection. Sci Rep. 4(1): https://doi.org/10.1038/srep04976
7.
Ye R, Xiang C, Lin J, Peng Z, Huang K, Yan Z, Cook NP, Samuel EL, Hwang C, Ruan G, et al. 2013. Coal as an abundant source of graphene quantum dots. Nat Commun. 4(1): https://doi.org/10.1038/ncomms3943
8.
Qu S, Wang X, Lu Q, Liu X, Wang L. 2012. A Biocompatible Fluorescent Ink Based on Water-Soluble Luminescent Carbon Nanodots. Angew. Chem.. 124(49):12381-12384. https://doi.org/10.1002/ange.201206791
9.
Wang D, Chen J, Dai L. 2015. Recent Advances in Graphene Quantum Dots for Fluorescence Bioimaging from Cells through Tissues to Animals. Part. Part. Syst. Charact.. 32(5):515-523. https://doi.org/10.1002/ppsc.201400219
10.
Kovalchuk A, Huang K, Xiang C, Martí AA, Tour JM. 2015. Luminescent Polymer Composite Films Containing Coal-Derived Graphene Quantum Dots. ACS Appl. Mater. Interfaces. 7(47):26063-26068. https://doi.org/10.1021/acsami.5b06057
11.
Meziani MJ, Dong X, Zhu L, Jones LP, LeCroy GE, Yang F, Wang S, Wang P, Zhao Y, Yang L, et al. 2016. Visible-Light-Activated Bactericidal Functions of Carbon ?Quantum? Dots. ACS Appl. Mater. Interfaces. 8(17):10761-10766. https://doi.org/10.1021/acsami.6b01765
12.
Zeng Z, Yu D, He Z, Liu J, Xiao F, Zhang Y, Wang R, Bhattacharyya D, Tan TTY. 2016. Graphene Oxide Quantum Dots Covalently Functionalized PVDF Membrane with Significantly-Enhanced Bactericidal and Antibiofouling Performances. Sci Rep. 6(1): https://doi.org/10.1038/srep20142
13.
Sun H, Gao N, Dong K, Ren J, Qu X. 2014. Graphene Quantum Dots-Band-Aids Used for Wound Disinfection. ACS Nano. 8(6):6202-6210. https://doi.org/10.1021/nn501640q
14.
Ting SL, Ee SJ, Ananthanarayanan A, Leong KC, Chen P. 2015. Graphene quantum dots functionalized gold nanoparticles for sensitive electrochemical detection of heavy metal ions. Electrochimica Acta. 1727-11. https://doi.org/10.1016/j.electacta.2015.01.026
15.
Sreeprasad TS, Rodriguez AA, Colston J, Graham A, Shishkin E, Pallem V, Berry V. 2013. Electron-Tunneling Modulation in Percolating Network of Graphene Quantum Dots: Fabrication, Phenomenological Understanding, and Humidity/Pressure Sensing Applications. Nano Lett.. 13(4):1757-1763. https://doi.org/10.1021/nl4003443
16.
Chao D, Zhu C, Zhang H, Shen ZX, Fan HJ. 2015. Graphene Quantum Dots Anchored VO2 Arrays to Boost the Electrochemical Performance of Li and Na Ion Batteries. https://doi.org/10.1364/oedi.2015.jw3a.22
17.
Sin Joo S, Kim J, Seok Kang S, Kim S, Choi S, Won Hwang S. 2014. Graphene-quantum-dot nonvolatile charge-trap flash memories. Nanotechnology. 25(25):255203. https://doi.org/10.1088/0957-4484/25/25/255203
18.
Guo C, Yang H, Sheng Z, Lu Z, Song Q, Li C. Layered Graphene/Quantum Dots for Photovoltaic Devices. Angewandte Chemie. 122(17):3078-3081. https://doi.org/10.1002/ange.200906291
19.
Chen Q, Wang C, Chen S. 2013. One-step synthesis of yellow-emitting carbogenic dots toward white light-emitting diodes. J Mater Sci. 48(6):2352-2357. https://doi.org/10.1007/s10853-012-7016-8