MilliporeSigma
HomeSterility TestingSteritest® NEO Membrane Filtration Sterility Test
Steritest® NEO Membrane Filtration Sterility Test
Latest Steritest® NEO Features
Sterility testing is one of the most crucial steps in pharmaceutical product release. The membrane filtration sterility test is the regulatory method of choice for filterable pharmaceutical products, as cited in the USP 71, Ph. Eur 2.6.1, and JP 4.06.
Steritest® Neo Devices
The test is particularly suitable for samples containing preservative, bacteriostatic, or fungistatic compounds, which inhibit the microbial growth of potential contaminants. With membrane filtration, microorganisms are retained by a 0.45 µm pore size filter, and all inhibiting compounds are rinsed using a suitable rinse solution. Large volumes and oily products dissolved in emulsifying agents can also be tested by the membrane filtration method, increasing sensitivity and reducing the volume of culture media. Appropriate culture media, selected based upon their ability to support the growth of anaerobic and aerobic microorganisms, are then transferred to the membrane filters. A 14-day incubation period is required to obtain final test results by checking turbidity.
The 4th generation of Steritest® devices is enriching with some additional elements contributing to significantly improving workflow safety, reliability, and convenience. Our sterility test consumables ensure that pharmaceutical products are never exposed to the environment during the testing process. Filtering, rinsing, media transferring, and incubating are all conducted within the Steritest® NEO closed system. There is no need to open the containers or manipulate the membrane at any time, thereby greatly reducing the risk of adventitious contamination resulting in false positives.
When used with the Steritest® Symbio pump and accessories, and Steritest® culture media and rinsing fluids, the Steritest® NEO devices offer the highest levels of quality and reliability and a fully regulatory compliant testing process.
Latest Steritest® NEO Features
The 4th generation of Steritest® devices with additional elements simplifies every aspect of testing, from handling to traceability to the reliability, all within a closed concept system. The ease and convenience enable you to increase productivity while maintaining the highest levels of quality.
Watch the video to discover all the new improvements.
New features of Steritest® NEO device is created to improve your workflow safety, reliability, and convenience.
Evolution of convenience
Color-coded protective cap in 2 parts help you to differentiate the needle type once covered.
Protective caps for long needles are now in 2 parts.
Watch the video to learn more
New 20 mm length needle is used to pierce cartridges or small soft plastic containers
Brand new short needle for small sample containers 
The pre-cut line in the accessory bag improves workflow convenience
An upgraded, easy-to-open accessory bag with a pre-cut line
Helps to precisely place the Steritest® tube in the pump head
New placement mark to place the tube in the pump head precisely
Cutting sterility testing tubes with a clamp
The new Velax® cutting clamp allows a safe and clean cut of the tubes
Pre-installed colored clamps help in accurate media filling
Pre-installed colored clamps prevent any media transfer errors 
Redesigned needle guard with grips
Redesigned needle guard with grips
The needle guard and protector improves confidence in needle manipulation
Newly designed needle protector with ridges
A needle being used with the Steritest® needle guide to access the sterility test sample in a Steritest® NEO canister.
The new needle guide allows safe and easy access to the sample anytime
Accessing a sterility test sample in a Steritest® NEO canister using the needle-free valve
The new needle-free valve is a safe solution for planned sampling or injection
Color-coded labels aid in packaging identification and selection of the appropriate box of Steritest® NEO device
Clear packaging identification with newly designed color-coded label based on the canister
Peel-and-stick label improves traceability
Peel-off label for improved traceability 
Smart 2D data matrix barcodes on labels for sterility testing
Smart Label with 2D data matrix barcode to retrieve product data for easy processing
Engraved 25 mL graduation line and volume for improved workflow accuracy
25 mL volume graduation marks on the canisters 
"Coloured clamps ensure ease of tracking tubing to canisters and make it easier to differentiate between media canisters, reducing the chance of wrong incubation.
The new tear bags are easy to open."
- Courtney Smith, Lead Technician - Microbiology Stockton Quality Control Laboratory - University Hospital of North Tees - United Kingdom
Meet the sterility testing industry benchmark
  • Minimize false positives: closed Steritest® NEO filtration devices reduce the risk of false positive results and avoid costly investigation and possible batch loss. There are no open containers or membrane manipulations, which could increase the risk of contamination.
  • Reduce false negatives: Steritest® NEO filtration devices are the right answer to the danger that false negative results pose to patients. Through specific membranes, unique sealing technology, and optimized device design, the unit allows the efficient elimination of bacteriostatic, fungistatic, or bactericidal agents.
  • Easy identification with color coding and optimized traceability.
  • Quality check: each Steritest® NEO device is subjected to rigorous in-process and releases quality checks, including 100% membrane and canister integrity tests as well as intense physical and microbiological testing. The detailed certificates of quality are available for download from our website
Get some insights in our high quality production of Steritest® NEO devices in our Center of Excellence, France.
Sterility testing solutions for products WITHOUT antimicrobial agents and medical devices
Steritest® NEO blue canisters for products without antimicrobial agents and medical devices
The “Blue Base” Steritest® NEO devices offer the ultimate flexibility. Perfect for use with most pharmaceutical drugs that do not have antimicrobial activity, our mixed esters of cellulose HA membrane allow fast flow rates for optimum throughput performance and reduction in testing time. A range of ergonomic needles is available to meet specific drug packaging requirements as well as simplify handling for gloved operators.
Liquids in ampoules and collapsible bags
Loading
Liquids in large vials
Loading
Liquids in small vials
Loading
Medical devices and collapsible bags
Loading
Liquids in syringes
Loading
Soluble powders in ampoules
Loading
Liquids in plastic containers
Loading
Soluble powders in vials
Loading
Sterility testing solutions for antibiotics, products WITH antimicrobial agents and medical devices
Steritest® NEO red canisters for antibiotics, products with antimicrobial agents, and medical devices
The “Red Base” Steritest® NEO devices, ideally suited for antibiotic sample testing, incorporate our Durapore® (PVDF) HV membrane, offering broad chemical compatibility and low binding properties. The chemical composition of the extremely thin 0.45 μm Durapore® membrane provides low antibiotic binding, which improves rinsing efficiency and reduces the risk of false negative results. The canister design ensures efficient rinsing of residual antimicrobial agents, and the needle and connections minimize the risk of antibiotic residuals
Antibiotics
Loading
Powders and super potent antibiotics
Loading
Liquids in large vial
Loading
Liquids in small vial
Loading
Soluble powders in vials
Loading
Medical devices and collapsible bags
Loading
Recommended accessories
Loading
Sterility testing solution for products dissolved in solvents requiring increased chemical compatibility
Steritest® NEO green canisters for products dissolved in solvents requiring increased chemical compatibility
The ‟Green Base” Steritest® NEO devices are suited for viscous products, such as creams and ointments, which are normally diluted in a sterile solvent, such as isopropyl myristate (IPM) to improve filterability. The solvent-resistant nylon canister and the Durapore® (PVDF) HV membrane ensure perfect chemical compatibility with IPM and other solvents. The reinforced base structure and canister connection guarantee an unmatched pressure resistance of the testing device during the whole filtration process. The ‟Green Base” Steritest® NEO devices are the perfect choice for testing solvents, creams, ointments, and veterinary injectables.
Loading
Sterility Testing Accessories for Liquid Transfer and Dilution
Streamline your workflow with our range of sterility testing accessories available for sample preparation and dilution. Steridilutor® NEO devices offers tubing and needle assembly to dissolve powders, or for the transfer of liquids. Steritest® vent needles is used for venting glass vials with rubber septa and rigid plastic vials, or media bottles.
Streamline your workflow with our range of sterility testing accessories available for sample preparation and dilution. Steridilutor® NEO devices offers tubing and needle assembly to dissolve powders, or for the transfer of liquids. Steritest® vent needles is used for venting glass vials with rubber septa and rigid plastic vials, or media bottles. The needles can also be used for sterility and growth promotion qualification of media batches.
Sample Preparation and Dilution
Loading
Liquid Transfer
Loading
Sterile Vent Needles
Loading
Sterility Test Sampling Solutions
Needle guide and needle-free valve sterility test sampling solutions mounted on Steritest® NEO canisters.
Throughout the incubation process of Steritest® NEO Devices for compendial sterility testing, it may be necessary to access the container to subculture, reincubate, or identify microorganisms within a turbid medium. It is pivotal to ensure that sterility remains intact and uncompromised throughout these procedures. Mounting a needle guide or a needle-free valve to your canister will help improve safety while performing this task. 
Loading
Complete Sterility Testing Solutions for Complete Confidence
Discover our full sterility testing portfolio based on over 50 years of experience and expertise. Our large variety of devices and pumps, along with sterile culture media and rinsing fluids can help you to stay compliant, whether you use membrane filtration or direct inoculation methods. Reduce the sterility testing workload and focus on your critical lab activities thanks to our extended Steritest® services portfolio. The experienced application and validation engineers will assist in method development and validation implementation within the QC microbiology laboratory. They will also provide basic and advanced technical training on sterility testing, in person or remotely.