MilliporeSigma
HomeSmall Molecules Analysis & QCFluticasone Propionate Assay by HPLC UV USP Monograph
Fluticasone Propionate Assay by HPLC-UV following USP Monograph
Eddy Tan, R&D Scientist
R&D APAC Lab (Singapore)
Chemical structure of fluticasone propionate
Fluticasone propionate
Abstract
The Purospher® STAR RP-18e Hibar® HPLC column is used for the assay of fluticasone propionate following the USP monograph. This is a reverse phase isocratic separation of fluticasone propionate from fluticasone propionate related compound D.
Introduction
Fluticasone propionate is a glucocorticoid and is available as nasal sprays to treat sneezing, itchy or runny nose, or other symptoms caused by hay fever. It is also available as creams and ointments for topical applications to treat itchy skin.
The Purospher® STAR RP-18e Hibar® HPLC column, 250 x 4.6 mm, 5 µm, is used to demonstrate the system suitability criteria for the assay of fluticasone propionate following the USP monograph.
Experimental
Mobile Phase Preparation
  • For buffer, weigh 1.15 g of monobasic ammonium phosphate in a 1 L volumetric flask and adjust with phosphoric acid to pH of 3.5 ± 0.05.
  • For mobile phase, make up methanol, acetonitrile and buffer as 50:15:35 parts respectively.
System Suitability Solution, 0.05 mg/mL USP Fluticasone Propionate Resolution Mixture RS
  • Weigh ~10 mg of USP Fluticasone Propionate Resolution Mixture RS into a 100 mL volumetric flask.
  • Add ~90 mL of mobile phase and sonicate for 15 min.
  • Top-up to mark with mobile phase and mix well. This is the System Suitability stock standard (~0.1 mg/mL).
  • Dilute to a concentration of 0.05 mg/mL with mobile phase.
Fluticasone Propionate Solutions for Assay and Linearity Test
  • Weigh ~10 mg of fluticasone propionate CRM into a 100 mL volumetric flask.
  •  Add ~90 mL of mobile phase to it and sonicate for 15 min.
  • Top-up to mark with mobile phase and mix well. This is the fluticasone propionate stock standard (~0.1 mg/mL).
  • Dilute to 0.01, 0.02, 0.03, 0.04, 0.05, 0.06, 0.07 mg/mL with mobile phase in 5 mL volumetric flasks.
Fluticasone propionate solution for assay and relative standard deviation (RSD): 0.04 mg/mL.
Fluticasone propionate solutions for linearity, LOD and LOQ: 0.01, 0.02, 0.03, 0.04, 0.05, 0.06, 0.07 mg/mL.
HPLC Parameters
Column:
Purospher® STAR RP-18e 5 µm Hibar® RT 250-4.6 mm (1.51456)
Detection:
UV, 239 nm
Mobile phase:
Methanol:acetonitrile:buffer (50:15:35), isocratic    
Flow rate:
1.5 mL/min
Pressure drop:
300 bar (or ~4350 psi)
Column temp.:
40 oC
Detector:
UV, 239 nm
Injection:
20 µL
Samples:
System Suitability Solution: 0.05 mg/mL USP Fluticasone Propionate Resolution Mixture RS in mobile phase (MP)
Fluticasone Propionate CRM solutions for assay and linearity test: 0.01, 0.02, 0.03, 0.04, 0.05, 0.06, and 0.07 mg/mL fluticasone propionate in MP
Results and Discussion
An HPLC-UV chromatogram, indicating the separation of fluticasone propionate and its related compound D. Two distinct  peaks, both colored green, are visible on the plot. The first peak corresponds to fluticasone propionate, observed at a retention time of 13.753 minutes, while the second peak represents fluticasone propionate related compound D, appearing at a retention time of 15.140 minutes.
Figure 1.0.05 mg/mL USP Fluticasone propionate resolution mixture RS.
The relative retention times for fluticasone propionate and fluticasone propionate related compound D are 1.00 and 1.10 respectively.
Injection number
Resolution (USP)
Injection 1
2.90
Injection 2
2.90
Injection 3
2.90
Injection 4
2.89
Injection 5
2.90
Average
2.90
Standard Deviation
0.004
RSD
0.15%
Table 1.Resolution between fluticasone propionate and related compound D for 0.05 mg/mL USP fluticasone propionate resolution mixture RS
Acceptance Criteria for System Suitability
Resolution: NLT 1.5 between fluticasone propionate and fluticasone propionate related compound D.
An HPLC-UV chromatogram illustrating the separation of fluticasone propionate. A singular green-colored peak is evident on the plot, corresponding to a concentration of 0.04 mg/mL of fluticasone propionate in the standard solution.
Figure 2.Fluticasone propionate 0.04 mg/mL standard solution.
Injection number
Peak Area
Injection 1
14.4618
Injection 2
14.4392
Injection 3
14.4089
Injection 4
14.4391
Injection 5
14.4354
Average
14.4369
Standard Deviation
0.019
RSD
0.13%
Table 2.Reproducibility of repeated injections of 0.04 mg/mL fluticasone propionate CRM solution
Acceptance Criteria for Standard Solution
Relative standard deviation (RSD): NMT 2%
Concentration (mg/mL)
Mean Area (mAU*min)
0.0100
3.581
0.0200
7.067
0.0300
10.773
0.0400
14.483
0.0500
17.877
0.0601
21.472
0.0701
25.231
STEYX
0.094
Slope
359.88
LOD (mg/mL)
0.00086
LOQ (mg/mL)
0.0026
R2
0.9999
Table 3.Linearity, LOD and LOQ for fluticasone propionate
A graphical plot, with peak area on the y-axis and concentration (mg/mL) on the x-axis, obtained for fluticasone propionate solutions ranging from 0.01 to 0.07 mg/mL. This graph demonstrates the method's linearity.
Figure 3.Linearity plot for fluticasone propionate solutions 0.01 to 0.07 mg/mL.
Conclusion
The Purospher® STAR RP-18e Hibar® RT column is able to comply with the system suitability criteria indicated in the USP monograph for the assay of fluticasone propionate. The resolution between fluticasone propionate and fluticasone propionate related compound D is greater than 1.5. The RSD of the standard solution is not more than 2%. Both criteria are within the assay specifications as defined in the monograph.
The column is robust with good reproducibility for repeated analyses (data not shown here). In addition, the column offers good linearity from 0.01 to 0.07 mg/mL fluticasone propionate with R2 of 0.9999. The Limit of Detection (LOD) for fluticasone propionate is 0.86 µg/mL and the Limit of Quantification (LOQ) is 2.6 µg/mL. 
Related Products
Loading
References
1.
USP Monographs, Fluticasone Propionate. USP-NF. Rockville, MD: United States Pharmacopeia. (2022). https://doi.org/10.31003/uspnf_m34220_05_01