70136
Deoxyribonuclease Test Agar
suitable for microbiology
Synonym(s):
DNase Test Agar
About This Item
Recommended Products
sterility
non-sterile
Quality Level
form
powder
shelf life
limited shelf life, expiry date on the label
composition
agar, 15 g/L
casein enzymic hydrolysate, 15 g/L
deoxyribonucleic acid, 2 g/L
papaic digest of soybean meal, 5 g/L
sodium chloride, 5 g/L
final pH
7.3±0.2 (25 °C)
application(s)
clinical testing
food and beverages
microbiology
suitability
nonselective and differential for Serratia spp.
nonselective and differential for Staphylococcus spp.
nonselective and differential for Streptococcus spp.
Application
Other Notes
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Chromogenic media enable the selective detection of S. aureus, which produce bluish-green colonies that are clearly differentiated from other species.
Streptococci- Overview of Detection, Identification, Differentiation and Cultivation Techniques
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service