MilliporeSigma
HomeMicrobial Cell CultureStreptococci - Overview of Detection, Identification, Differentiation and Cultivation Techniques
Streptococci - Overview of Detection, Identification, Differentiation and Cultivation Techniques
Jvo Siegrist
AnalytiX Volume 7 Article 3
Streptococci are non-motile, microaerophilic, Grampositive spherical bacteria (cocci). They often occur as chains or pairs and are facultative or strict anaerobes. Streptococci give a negative catalase test, while staphylococci are catalase-positive. The cell division of Streptococci species involves two separate biosynthetic events: peripheral cell-wall elongation and septal-wall synthesis.1 Because they are unable to synthesize cytochromes, Streptococci cannot carry out oxidative phosphorylation. They are able to ferment sugars, but the end product is always lactic acid. Therefore, Streptococci are very acid tolerant and count among the lactic acid bacteria order.
There are many natural sources of Streptococci, including humans and diverse animals where they often colonize the mucosal surfaces of mouth, intestinal tract, nasal passages and pharynx. The presence of Streptococci  in drinking water indicates fecal contamination. Food sources with high risk of Streptococcal contamination include milk and dairy products, eggs, steamed lobster, ground ham, potato salad, custard, rice pudding and shrimp salad. In most Streptococcal food poisoning cases, the food was allowed to stand at room temperature for several hours between preparation and consumption. The contamination of the food is most often the result of poor hygiene, handling of the food by infected people, or the use of raw (unpasteurized) milk. Although they can be potent pathogens, some Streptococci are commercially important for the production of cheese and yogurt. These include S. lactis, S. cremoris, S. diacelillactis and S. thermophilus, the latter being the most well-known.
For detection, identification, differentiation, enumeration and cultivation of Streptococci, we provide a broad range of specific agars and broths (Table 1), Streptococci Diagnostic Tests (Table 2) and a Gram staining kit and component solutions (Table 3).
Streptococci
Scientific classification of Streptococcus
Kingdom: Eubacteria
Phylum: Firmicutes
Class: Bacilli
Order: Lactobacillales
Family: Streptococcaceae
Genus: Streptococcus
Grouping of Streptococci based on hemolytic activity
Streptococci are divided into three groups based on their hemolytic (red blood cell lysing) activity. The hemolytic reaction can be visualized on blood agar plates, such as the non-selective Agars for Differentiation that are listed in Table 1.
Non-selective Enrichment Broths
Cat. No.BrandDescription
53286Sigma-AldrichBrain Heart Broth
B2551Sigma-AldrichBrewer thioglycollate medium
22089Sigma-AldrichCasein peptone Lecithin Polysorbate Broth
22098Sigma-AldrichCASO Broth
60865Sigma-AldrichCooked Meat Broth
17123Sigma-AldrichElliker Broth
5121Sigma-AldrichHeart Infusion Broth
70122Sigma-AldrichNutrient Broth No. 1
70149Sigma-AldrichNutrient Broth No. 3
40893Sigma-AldrichPeptone Water, phosphate-buffered, Vegitone
70179Sigma-AldrichPeptone Water
77187Sigma-AldrichPeptone Water, phosphate-buffered
S4681Sigma-AldrichStandard Nutrient Broth No. 1
85905Sigma-AldrichStuart Ringertz Medium
90404Sigma-AldrichThioglycolate Broth with Resazurine
T1438Sigma-AldrichTodd Hewitt Broth
22092Sigma-AldrichTryptic Soy Broth
51228Sigma-AldrichTryptic Soy Broth No. 2
41298Sigma-AldrichTryptic Soy Broth, Vegitone
T3938Sigma-AldrichTryptone Soya Broth without Dextrose
T4407Sigma-AldrichTryptose Broth
V5262Sigma-AldrichVeal Infusion Broth
Selective Enrichment Broths
Cat. No.BrandDescription
17157Sigma-AldrichGlucose Azide Broth
Non-selective Agars for Cultivation, Enumeration and Isolation
Cat. No.BrandDescription
A3340Sigma-AldrichAC Agar
70138Sigma-AldrichBrain Heart Infusion Agar
22095Sigma-AldrichCASO Agar
27688Sigma-AldrichColumbia Agar
D3060Sigma-AldrichDextrose starch agar
D8184Sigma-AldrichDiagnostic sensitivity test agar
70147Sigma-AldrichMilk Agar
70148Sigma-AldrichNutrient Agar
17197Sigma-AldrichPeptonized Milk Agar
70152Sigma-AldrichPlate Count Agar
88588Sigma-AldrichPlate Count Agar according to Buchbinder et al.
19718Sigma-AldrichPlate Count Agar, Vegitone
17257Sigma-AldrichStreptococcus thermophilus Isolation Agar
22091Sigma-AldrichTryptic Soy Agar
14432Sigma-AldrichTryptic Soy Agar, Vegitone
51414Sigma-AldrichTryptic Soya Agar with Polysorbate 80 and Lecithin
70159Sigma-AldrichTryptone Glucose Extract Agar
T2313Sigma-AldrichTryptose Agar
Non-selective Agars for Differentiation
Cat. No.BrandDescription
70133Sigma-AldrichBlood Agar (Base)
B1676Sigma-AldrichBlood Agar Base No. 2
55420Sigma-AldrichCLED Agar
70136Sigma-AldrichDeoxyribonuclease Test Agar
D2560Sigma-AldrichDNase Test Agar with Toluidine Blue
17153Sigma-AldrichLS Differential Agar
Selective Agars with Differential System for Differentiation, Detection and Isolation
Cat. No.BrandDescription
06105Sigma-AldrichBile Esculin Azide Agar
17151Sigma-AldrichKanamycin Esculin Azide Agar
01337Sigma-AldrichMitis Salivarius Agar
Media for Sensitivity Testing
Cat. No.BrandDescription
97580Sigma-AldrichMueller-Hinton Agar 2
Table 1Media for Streptococci
Alpha-Hemolytic Streptococci
In alpha hemolysis, the red blood cells remain intact, but the hemoglobin is converted to biliverdin. This causes dark greening of the blood agar plate around the colonies.
  • Pneumococci
    S. pneumoniae: Causes bacterial pneumonia, otitis media and meningitis. S. pneumoniae sensitivity to optochin (ethyl hydrocupreine hydrochloride) is the basis of a diagnostic test (Figure 1, Table 2).
Pneumococci
Figure 1.Differentiation and identification flow chart of Gram-positive cocci
Cat. No.BrandDescription
88597Sigma-AldrichCatalase Test
08382Sigma-AldrichBacitracin Disks
74042Sigma-AldrichOptochin Disks
67886Sigma-AldrichPYRase Strips
Table 2Diagnostic tests for identification and differentiation of streptococci
  • Viridans and Others
    S. mutans: Associated with dental caries.
    S. viridans: Causative agent for endocarditis and dental abscesses.
    S. salivarius: Considered to be an opportunistic pathogen.
    S. salivarius subsp. thermophilus: Used in the production of cheese and yogurt.
Beta-Hemolytic Streptococci
Beta hemolysis is a complete hemolysis of erythrocytes by the enzyme hemolysin. Clear zones will appear around the colonies on the blood agar plate. Beta-hemolytic Streptococci are further divided into serological groups using specific antibodies that recognize surface carbohydrate antigens (e.g. A, B, etc.).
  • Group A
    S. pyogenes: Causes infections like strep throat, acute rheumatic fever, scarlet fever, acute glomerulonephritis and necrotizing fasciitis. It is sensitive to bacitracin (Fig. 1), a fact that is employed in a Streptococcus diagnostic test (Table 2). Other Streptococci species may also possess the Group A antigen but are not human pathogens. These include S. dysgalactiae subsp. equisimilis and S. anginosus strains.
  • Group B
    S. agalactiae: Causes meningitis in neonates and the elderly. Occasionally colonizing the female reproductive tract, they increase the risk for premature rupture of membranes and transmission of the infection to the child.
  • Group C
    S. equi: Causes strangles infection in horses.
    S. zooepidemicus: Causes infections in cattle, horses and other mammals.
  • Group D (Enterococci)
    Many former Group D Streptococci have been reclassified to the genus Enterococcus (e.g. S. faecalis, S. faciem, S. durans, S avium).
    S. bovis and S. suis: Still in the Group D streptococci.
Non-Hemolytic or Gamma-Hemolytic Streptococci
Gamma hemolysis is a misnomer as there is actually no hemolysis. Non-hemolytic or gamma-hemolytic Streptococci rarely cause disease.
Cat. No.BrandDescription
77730Sigma-AldrichGram Staining Kit
94448Sigma-AldrichGram‘s Crystal violet Solution
75482Sigma-AldrichGram‘s Decolorizer Solution
87794Sigma-AldrichGram’s Fuchsin Solution
90107Sigma-AldrichGram‘s Iodine Solution
94635Sigma-AldrichGram‘s Safranin Solution
Table 3Gram staining kit and component solutions
Materials
Loading
References
1.
Beilharz K, Novakova L, Fadda D, Branny P, Massidda O, Veening J. 2012. Control of cell division in Streptococcus pneumoniae by the conserved Ser/Thr protein kinase StkP. Proceedings of the National Academy of Sciences. 109(15):E905-E913. https://doi.org/10.1073/pnas.1119172109