Skip to Content
Merck
All Photos(1)

Key Documents

SRP4754

Sigma-Aldrich

WNT-1 human

recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)

Synonym(s):

INT-1, WNT1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

potency

1.5-2.5 ng/mL ED50

mol wt

~38.4 kDa

packaging

pkg of 10 μg

storage condition

avoid repeated freeze/thaw cycles

impurities

endotoxin, tested

NCBI accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... WNT1(7471)

General description

Wnt family member 1 (Wnt-1) is a secreted protein that signals through the Frizzled family of cell surface receptors. Mature human Wnt-1 is a glycosylated protein containing 343 amino acid residues. The gene encoding this protein is localized on human chromosome 12q13.12. Recombinant human Wnt-1 is a 38.4kDa, non-glycosylated protein containing 343 amino acid residues.

Application

Wnt family member 1 (Wnt-1) has been used in the differentiation medium for human neural progenitor cells (hNP1).

Biochem/physiol Actions

Wnt family member 1 (Wnt-1) and is required for normal embryonic development. Wnt-1 activation induces a complex signaling cascade that ultimately leads to the increased expression of over 50 genes. An important component of Wnt-1 signaling is the stabilization, and resulting accumulation, of the intracellular signaling protein, β-catenine. Wnt signaling induces and maintains the transformed phenotype and, in certain embryonic cell lines, supports self-renewal in the absence of significant differentiation. Elevated levels of Wnt proteins are associated with tumorigenesis and are present in numerous human breast cancers.

Physical form

Sterile filtered and lyophilized with no additives.

Reconstitution

Centrifuge the vial prior to opening. Avoid freeze-thaw cycles.
Reconstitute in water to a concentration of 0.1-1.0 μg/μL. The solution can then be diluted into other aqueous buffers.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Derivation of sensory neurons and neural crest stem cells from human neural progenitor hNP1.
Guo X
Biomaterials, 34(18), 4418-4427 (2013)
Promoting human embryonic stem cell renewal or differentiation by modulating Wnt signal and culture conditions.
Cai L
Cell Research, 17(1), 62-72 (2007)
Cross-talk of WNT and FGF signaling pathways at GSK3beta to regulate beta-catenin and SNAIL signaling cascades.
Katoh M and Katoh M
Cancer Biology & Therapy, 5(9), 1059-1064 (2006)
Wnt/beta-catenin signaling: components, mechanisms, and diseases.
MacDonald BT
Developmental Cell, 17(1), 9-26 (2009)
Xue Hong et al.
Frontiers in physiology, 12, 745816-745816 (2021-11-26)
Studies suggest that Wnt/β-catenin agonists are beneficial in the treatment of acute kidney injury (AKI); however, it remains elusive about its role in the prevention of AKI and its progression to chronic kidney disease (CKD). In this study, renal Wnt/β-catenin

Articles

Role of growth factors in stem cell differentiation and various growth factors for your research at sigmaaldrich.com

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service