コンテンツへスキップ
Merck
  • Transgenerational inheritance of centromere identity requires the CENP-A N-terminal tail in the C. elegans maternal germ line.
Transgenerational inheritance of centromere identity requires the CENP-A N-terminal tail in the C. elegans maternal germ line.
PLoS biology (2021-07-07)
Reinier F Prosée, Joanna M Wenda, Isa Özdemir, Caroline Gabus, Kamila Delaney, Francoise Schwager, Monica Gotta, Florian A Steiner
要旨
Centromere protein A (CENP-A) is a histone H3 variant that defines centromeric chromatin and is essential for centromere function. In most eukaryotes, CENP-A-containing chromatin is epigenetically maintained, and centromere identity is inherited from one cell cycle to the next. In the germ line of the holocentric nematode Caenorhabditis elegans, this inheritance cycle is disrupted. CENP-A is removed at the mitosis-to-meiosis transition and is reestablished on chromatin during diplotene of meiosis I. Here, we show that the N-terminal tail of CENP-A is required for the de novo establishment of centromeres, but then its presence becomes dispensable for centromere maintenance during development. Worms homozygous for a CENP-A tail deletion maintain functional centromeres during development but give rise to inviable offspring because they fail to reestablish centromeres in the maternal germ line. We identify the N-terminal tail of CENP-A as a critical domain for the interaction with the conserved kinetochore protein KNL-2 and argue that this interaction plays an important role in setting centromere identity in the germ line. We conclude that centromere establishment and maintenance are functionally distinct in C. elegans.
材料
製品番号
ブランド
製品内容
Sigma-Aldrich
モノクロナール抗α-チューブリン マウス宿主抗体, clone DM1A, ascites fluid
Millipore
グルタチオンHigh Capacity磁気アガロースビーズ, (1:1 suspension in a 30% ethanol solution)