HPLCによるエネルギードリンクの分析、Ascentis Express RP-AmideおよびUV検出使用
材料
Standard
分析カラム
column
Ascentis Express RP-Amide, 5 cm x 3 mm I.D., 2.7 μm particles (53916-U)
mobile phase
[A] water with 0.1% TFA; [B] acetonitrile with 0.1% TFA
flow rate
0.6 mL/min
pressure
1146 psi (79 bar)
column temp.
35 °C
detector
UV, 254 nm
injection
2 μL
sample
Energy Drink Diluted 1:9 in acetonitrile
詳細
アナリシスノート
This application demonstrates the suitability of Ascentis Express RP-Amide for the efficient separation of common drink additives used in caffeinated energy drinks.