HPLCによるメチオニンエナンチオマーの分析、CHIROBIOTIC® T2
CONDITIONS
column
CHIROBIOTIC T2, 25 cm x 4.6 mm I.D., 5 μm particles (16024AST)
mobile phase
[A] water; [B] acetonitrile; (25:75, A:B)
flow rate
1 mL/min
column temp.
25 °C
detector
UV, 220 nm