주요 문서
ESYNTH008
Sigma-Aldrich
SynLectro Electrode Holder
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
81141502
NACRES:
NB.61
일반 설명
SynLectro Electrode Holder is part of the SynLectro Electrolysis Platform for synthetic electrochemistry. This standard electrode holder is designed to hold electrodes in firmly place to create a homogenous electric field.
SynLectro Electrolysis Platform
The SynLectro Electrolysis Platform is a portfolio of labware products and electrodes designed to simplify and standardize preparative scale electrosynthesis. Learn more about each component, designed with consistency, reproducibility, and safety in mind.
법적 정보
SynLectro is a trademark of Merck KGaA, Darmstadt, Germany
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)
문서 라이브러리 방문
관련 콘텐츠
Learn about our new SynLectro™ Electrolysis Platform that has been designed to simplify and standardize electrochemical organic synthesis at research and batch scale.