문서
Z190403
Sigma-Aldrich
Aldrich® flask-type sprayer
Replacement Head only
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41121800
NACRES:
NB.51
material
borosilicate glass
무균
non-sterile
특징
closure type Rubber cork
포장
pack of 1
일반 설명
Connect using the barb end to a low-pressure gas or air source of <5 psi. By positioning your thumb over the vent hole you can control the spray pattern.
법적 정보
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC