주요 문서
55053
RheBuild® Kit
for use with 9725/9725i/9726 Injectors
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41115854
호환성
for use with 9725/9725i/9726 Injectors
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
These kits include all of the tools and parts you need to repair your Rheodyne injector. The parts included are those most likely to be damaged, worn, or lost. Kits for front-loading injectors include: rotor seal, stator face assembly, isolation seal, needle guide, needle port cleaner, 2 hex keys, operating instructions, and a mini-manual. Parts may vary, depending on valve model.
추천 제품
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
법적 정보
RheBuild is a registered trademark of IDEX Health & Science, LLC
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)