콘텐츠로 건너뛰기
Merck
  • Adverse morphological development in embryonic zebrafish exposed to environmental concentrations of contaminants individually and in mixture.
Adverse morphological development in embryonic zebrafish exposed to environmental concentrations of contaminants individually and in mixture.
Aquatic toxicology (Amsterdam, Netherlands) (2016-04-24)
Cassandra D Kinch, Deborah M Kurrasch, Hamid R Habibi
초록
Exposure to environmental contaminants has been linked to developmental and reproductive abnormalities leading to infertility, spontaneous abortion, reduced number of offspring, and metabolic disorders. In addition, there is evidence linking environmental contaminants and endocrine disruption to abnormal developmental rate, defects in heart and eye morphology, and alterations in behavior. Notably, these effects could not be explained by interaction with a single hormone receptor. Here, using a whole-organism approach, we investigated morphological changes to developing zebrafish caused by exposure to a number of environmental contaminants, including bisphenol A (BPA), di(2-ethylhexyl)phthalate (DEHP), nonylphenol, and fucosterol at concentrations measured in a local water body (Oldman River, AB), individually and in mixture. Exposure to nanomolar contaminant concentrations resulted in abnormal morphological development, including changes to body length, pericardia (heart), and the head. We also characterize the spatiotemporal expression profiles of estrogen, androgen, and thyroid hormone receptors to demonstrate that localization of these receptors might be mediating contaminant effects on development. Finally, we examined the effects of contaminants singly and in mixture. Combined, our results support the hypothesis that adverse effects of contaminants are not mediated by single hormone receptor signaling, and adversity of contaminants in mixture could not be predicted by simple additive effect of contaminants. The findings provide a framework for better understanding of developmental toxicity of environmental contaminants in zebrafish and other vertebrate species.
MATERIALS
제품 번호
브랜드
제품 설명
Sigma-Aldrich
TWEEN® 20, viscous liquid, suitable for cell culture
로그인조직 및 계약 가격 보기
SKU팩 크기재고 정보가격수량
250 mL
예상 입고일 2025년 1월 20일
세부사항...
₩62,626
Sigma-Aldrich
Fucosterol, ≥93%
로그인조직 및 계약 가격 보기
SKU팩 크기재고 정보가격수량
250 mL
예상 입고일 2025년 1월 20일
세부사항...
₩62,626