Skip to Content
Merck
All Photos(1)

Key Documents

90035

Millipore

DCLS Agar No. 2

suitable for microbiology

Synonym(s):

Desoxycholate Citrate Lactose Sucrose Agar No. 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171606

form

powder

composition

agar, 12.0 g/L
beef extract, 3.0 g/L
lactose, 5.0 g/L
neutral red, 0.03 g/L
proteose peptone, 7.0 g/L
saccharose, 5.0 g/L
sodium citrate, 10.0 g/L
sodium deoxycholate, 2.5 g/L
sodium thiosulphate, 5.0 g/L

final pH

7.2±0.2 (25 °C)

application(s)

clinical testing
environmental
food and beverages

microbiology

suitability

selective and differential for Enterobacter spp.
selective and differential for Proteus spp.
selective and differential for Salmonella spp.
selective and differential for Serratia spp.
selective and differential for Shigella spp.
selective and differential for Vibrio spp.
selective and differential for Yersinia spp.

Application

D.C.L.S. Agar is used as a selective medium for the isolation of Shigella and Salmonella. Also useful for isolation of Vibrio cholera. The addition of fermentable carbohydrate increases the usefulness of the medium because non-pathogenic sucrose-fermenting organisms may be recognised by their red colonies, e.g. some Proteus, Serratia, Enterobacter species.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service