Skip to Content
Merck
All Photos(1)

Key Documents

712604

Sigma-Aldrich

O-(2-Azidoethyl)-O′-methyl-undecaethylene glycol

≥90% (NMR)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H51N3O12
Molecular Weight:
585.69
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Pricing and availability is not currently available.

assay

≥90% (NMR)

form

liquid

reaction suitability

reaction type: click chemistry
reagent type: cross-linking reagent

storage temp.

2-8°C

SMILES string

COCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCN=[N+]=[N-]

InChI

1S/C25H51N3O12/c1-29-4-5-31-8-9-33-12-13-35-16-17-37-20-21-39-24-25-40-23-22-38-19-18-36-15-14-34-11-10-32-7-6-30-3-2-27-28-26/h2-25H2,1H3

InChI key

FXVJBKLBTILMNA-UHFFFAOYSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Articles

Circulatory half-life is a key success factor for new drugs. In this respect, PEGylation or PEG-ing—the modification of potential candidates ranging from non-peptidic small molecules to peptides and proteins, antibody fragments, aptamers, and saccharides or oligonucleotides with polyethylene glycol chains—offers numerous advantages.

Prof. Randal Lee discusses iron oxide magnetic nanospheres and nanocubes design considerations for biosensing applications.

Kanjiro Miyata (The University of Tokyo, Japan) provides insights on the rational design of polymeric materials for “smart” oligonucleotide delivery.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service