Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CDS019936

Sigma-Aldrich

Isoflurane

AldrichCPR

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C3H2ClF5O
CAS Number:
Molecular Weight:
184.49
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Looking for similar products? Visit Product Comparison Guide

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

STOT SE 3

target_organs

Central nervous system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

  1. Which document(s) contains shelf-life or expiration date information for a given product?

    If available for a given product, the recommended re-test date or the expiration date can be found on the Certificate of Analysis.

  2. How do I get lot-specific information or a Certificate of Analysis?

    The lot specific COA document can be found by entering the lot number above under the "Documents" section.

  3. How do I find price and availability?

    There are several ways to find pricing and availability for our products. Once you log onto our website, you will find the price and availability displayed on the product detail page. You can contact any of our Customer Sales and Service offices to receive a quote.  USA customers:  1-800-325-3010 or view local office numbers.

  4. What is the Department of Transportation shipping information for this product?

    Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product. 

  5. What are the density, volume and concentration of Product CDS019936, Isoflurane?

    Based on a density of 1.496 g/mL, 250 milligrams equals 167 microliters. Given a molecular weight of 184.5 grams per mole, the neat liquid has an effective concentration of 8.1 M.

  6. My question is not addressed here, how can I contact Technical Service for assistance?

    Ask a Scientist here.

Yanlin Jia et al.
Nature communications, 5, 4387-4387 (2014-07-19)
Injury to lung epithelial cells has a role in multiple lung diseases. We previously identified mitsugumin 53 (MG53) as a component of the cell membrane repair machinery in striated muscle cells. Here we show that MG53 also has a physiological
Oliver Grottke et al.
Critical care (London, England), 18(1), R27-R27 (2014-02-07)
New oral anticoagulants are effective alternatives to warfarin. However, no specific reversal agents are available for life-threatening bleeding or emergency surgery. Using a porcine model of trauma, this study assessed the ability of prothrombin complex concentrate (PCC), activated PCC (aPCC)
Romain Sonneville et al.
Shock (Augusta, Ga.), 44(3), 245-251 (2015-05-27)
Glucose toxicity may play a crucial role in evoking neurologic complications of critical illness. We studied whether the neuropathological alterations in fatal human critical illness observed under hyperglycemia are present and can be attenuated by maintaining normoglycemia in a mouse
Daniela S Ardelean et al.
Angiogenesis, 17(3), 641-659 (2014-02-11)
Chronic intestinal inflammation is associated with pathological angiogenesis that further amplifies the inflammatory response. Vascular endothelial growth factor (VEGF), is a major angiogenic cytokine that has been implicated in chronic colitis and inflammatory bowel diseases. Endoglin (CD105), a transforming growth
David A X Nayagam et al.
PloS one, 9(5), e97182-e97182 (2014-05-24)
To assess the safety and efficacy of chronic electrical stimulation of the retina with a suprachoroidal visual prosthesis. Seven normally-sighted feline subjects were implanted for 96-143 days with a suprachoroidal electrode array and six were chronically stimulated for 70-105 days

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service