Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D-109

Supelco

Dihydrocodeine-6ß-D-glucuronide solution

1.0 mg/mL in methanol: water (1:1), certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H31NO9
CAS Number:
Molecular Weight:
477.50
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Decreto Lei 15/93: Tabela IA (Portugal)

concentration

1.0 mg/mL in methanol: water (1:1)

format

single component solution

storage temp.

−20°C

SMILES string

O[C@H]1[C@H](O[C@@H](CC2)[C@H]3[C@]([C@]2([H])[C@H]4C5)(CCN4C)C6=C5C=CC(OC)=C6O3)O[C@H](C(O)=O)[C@@H](O)[C@@H]1O

InChI

1S/C24H31NO9/c1-25-8-7-24-11-4-6-14(32-23-18(28)16(26)17(27)20(34-23)22(29)30)21(24)33-19-13(31-2)5-3-10(15(19)24)9-12(11)25/h3,5,11-12,14,16-18,20-21,23,26-28H,4,6-9H2,1-2H3,(H,29,30)/t11-,12+,14-,16-,17-,18+,20-,21-,23+,24-/m0/s1

InChI key

WBVBRJPPWYRNPU-HSCJLHHPSA-N

General description

Dihydrocodeine-6β-D-glucuronide is a major urinary metabolite of dihydrocodeine, an opiate used for treatment of moderate to severe pain and as an antitussive. Dihydrocodeine is twice as potent as codeine and is frequently abused in Europe and in Asia. This Certified Spiking Solution® is suitable for use as a starting material in calibrators and controls for LC/MS and GC/MS testing methods in clinical toxicology, forensic analysis, pharmaceutical research, and pain prescription monitoring.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 3 - Flam. Liq. 3 - STOT SE 1

target_organs

Eyes

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

75.2 °F

flash_point_c

24 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service