Key Documents
1.67710
Supelco
Continous level sensor for organic solvents in 10l stainless steel barrels
About This Item
UNSPSC Code:
41122409
NACRES:
NA.03
storage temp.
no temp limit
Related Categories
General description
Continuous level sensor technology with ATEX approval provides maximum safety and reliability for your daily work. The graphical display always shows the exact solvent level inside the container. With adjustable alarm, individual management, measurement and control of solvent supply and disposal now are available and automatable, e.g. by visible control alarms or automatical control of HPCL instruments.
incl. G2 threaded adapter, dip tube, graphical display and cables (length: 5m)