Skip to Content
Merck
All Photos(1)

Key Documents

654164

Sigma-Aldrich

Tubulin Polymerization Inhibitor II

The Tubulin Polymerization Inhibitor II, also referenced under CAS 1151995-69-5, acts as an effective anti-microtubule agent. This small molecule/inhibitor is primarily used for Cell Structure applications.

Synonym(s):

Tubulin Polymerization Inhibitor II, (E)-1,3-Dihydro-6-methoxy-3-(3,4,5-trimethoxybenzylidene)-1H-indol-2-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H19NO5
CAS Number:
Molecular Weight:
341.36
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77

Pricing and availability is not currently available.

Quality Level

assay

≥97% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 10 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

N1c2c(ccc(c2)OC)C(=Cc3cc(c(c(c3)OC)OC)OC)C1=O

General description

A cell-permeable, SU5416- (Cat. Nos. 676487 and 676498) derived combretastatin A-4 (Cat. No. 234200) analog that acts as an effective anti-microtubule agent (IC50 = 4.5 µM in inhibiting polymerization of purified porcine brain tubulin) and displays extremely potent anti-proliferative activity towards various cancer cell lines (GI50<10 nM for 46 lines among a 53 NCI panel tested).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Purified porcine brain tubulin
Product does not compete with ATP.
Reversible: no
Target IC50: 4.5 µM in inhibiting polymerization of purified porcine brain tubulin

Packaging

Packaged under inert gas

Warning

Toxicity: Harmful (C)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Pandit, B., et al. 2006. Bioorg. Med. Chem.14, 6492.
Li, P.K., et al. 2005. Bioorg. Med. Chem. Lett.15, 5382.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service