Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MAB1098

Sigma-Aldrich

Anti-Tumor Necrosis Factor-β Antibody, clone 9B9

clone 9B9, Chemicon®, from mouse

Synonym(s):

TNF-beta

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

9B9, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... LTA(4049)

Specificity

This antibody is use for detection, identification and neutralixzation of TNF-b in cell culture and other biological samples. The antibody reacts with human natural and recombinant TNF-b (lymphotoxin). It does not react with human TNF-a. Determination of cross-reactivity to TNF-b from other species has not been done.

Immunogen

Human, recombinant tumor necrosis factor-beta (TNF-beta, lymphotoxin) produced in E. Coli.

application

Anti-Tumor Necrosis Factor-β Antibody, clone 9B9 detects level of Tumor Necrosis Factor-β & has been published & validated for use in IC, IH & WB.
Western blot

Immunocytochemistry

Immunohistochemistry

Neutralization: 200ng of TNF-beta antibody is required for the neutralizaion of 1 unit/ml TNF-beta. The TNF-beta-preparation which is used has a specific activity of ca. 2x10(7) units/mg protein [cell lytic assay with L 929 cells (mouse transformed fibroblasts) and/or WEHJ 164 cells (mouse fibrosarcoma cells)/actinomycin D] .

Optimal working dilutions must be determined by end user.

Physical form

Format: Purified
Purified immunoglobulin (Protein A-agarose chormotography). One bottle of lyophilized preparation contains:200ug mouse IgG, 2 mg potassium phosphate buffer, 2.9 mg sodium chloride, and 20 mg raffinose.Reconstitution with 1 ml sterile, distilled water results in an antibody concentration of 200 ug/ml.

Storage and Stability

Maintain frozen at -20°C in undiluted aliquots for up to 12 months.Freeze only once.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service