Key Documents
SPKGENINS037
Millipore
Spare part, Conductivity Sensor
0 - 850 mS/cm
About This Item
UNSPSC Code:
23151800
material
PEEK
Quality Level
product line
Mobius®
compatibility
for use with Mobius®
General description
The conductivity sensor is an analytical device used to measure the electrical conductivity of aqueous solutions. This sensor is important for monitoring and ensuring the quality pharmaceutical applications.
This spare part is classified as ′Critical,′ meaning that it is crucial, for avoiding system downtime, to keep at least one in stock for immediate availability.
Packaging
Pack of 1 unit
Legal Information
MOBIUS is a registered trademark of Merck KGaA, Darmstadt, Germany