Key Documents
SPKMBCHRAUT002
Millipore
Spare part, Mobius® Chrom, Cube67 Bus Node
Clamshell for Mobius® Chrom 10
About This Item
UNSPSC Code:
23151800
Pricing and availability is not currently available.
material
PBT
Quality Level
product line
Mobius®
compatibility
for use with Mobius®
General description
This Bus Node, designed for Mobius® Chrom systems, serves as signal communication within the equipment.
This spare part is classified as ′Critical,′ meaning that it is crucial, for avoiding system downtime, to keep at least one in stock for immediate availability.
Packaging
Pack of 1 unit
Legal Information
MOBIUS is a registered trademark of Merck KGaA, Darmstadt, Germany