Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

V311-15GS

Tryptic Soy Broth- Ready-to-use Bags, Tubes & Ampoules

vial capacity × filling volume 20 mL × 15 mL, closure type, blue aluminum rubber septum, box of 25 vials, suitable for sterility testing

Synonym(s):

CASO Broth, Casein Soy Broth, Casein Soyabean Digest Broth, TSB, Tryptic Soy Broth (TSB), Tryptone Soy Broth

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106214

material

glass vial

Quality Level

agency

EP 2.6.13
JP 4.06
USP 71

sterilization compatibility

autoclavable compatible

form

liquid

feature

closure type blue aluminum rubber septum
ready-to-use

packaging

box of 25 vials

technique(s)

microbiological culture: suitable
sterility testing: suitable

vial capacity × filling volume

20 mL × 15 mL

color

amber

pH

7.3±0.2 (25 °C)

application(s)

food and beverages
pharmaceutical

storage temp.

2-25°C

suitability

nonselective for

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Tryptic Soy Broth is a nutrient-rich universal medium free of inhibitors and targets to enumerate a broad spectrum of bacteria, yeast, and molds. The medium is supplemented with peptones from casein and soymeal to provide an enhanced carbon source, which promotes the growth of the microflora. The broth medium is used during sterility testing due to its wide range of specificity.

Application

Tryptic Soy broth is used to cultivate and enumerate a broad range of bacteria, yeast and fungi present in a test sample.

pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Resp. Sens. 1

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service