53605
Violet Red Bile Glucose Agar without Lactose, Vegitone
suitable for microbiology
Synonym(s):
VRBG Agar without Lactose, Vegitone, Vegitone Violet Red Bile Glucose Agar without Lactose, Violet Red Bile Dextrose Agar without Lactose, Vegitone
About This Item
Recommended Products
shelf life
limited shelf life, expiry date on the label
Quality Level
composition
agar, 12.0 g/L
crystal violet, 0.002 g/L
glucose, 10.0 g/L
neutral red, 0.03 g/L
peptone (vegetable), 7.0 g/L
sodium chloride, 5.0 g/L
synthetic detergent (bile salt replacement), 1.5 g/L
yeast extract, 3.0 g/L
final pH
7.4±0.2 (25 °C)
application(s)
agriculture
environmental
food and beverages
microbiology
suitability
selective and differential for Escherichia coli
selective and differential for Salmonella spp.
selective and differential for Shigella spp.
selective and differential for Yersinia spp.
selective and differential for coliforms
selective and differential for enterobacteriaceae
Application
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service