92322
Rappaport Vassiliadis medium (base), modified, semi-solid
suitable for microbiology, NutriSelect® Plus
Synonym(s):
MSRV medium (base), Modified Semisolid Rappaport Vassiliadis Medium (base)
About This Item
Recommended Products
sterility
non-sterile
Quality Level
form
powder
shelf life
limited shelf life, expiry date on the label
composition
agar, 2.7 g/L
casein peptone, 4.59 g/L
magnesium chloride, 10.93 g/L
malachite green, 0.037 g/L
mono-potassium phosphate, 1.47 g/L
sodium chloride, 7.34 g/L
tryptose, 4.59 g/L
manufacturer/tradename
NutriSelect® Plus
technique(s)
microbiological culture: suitable
final pH
5.2±0.2 (25 °C)
application(s)
environmental
food and beverages
sample preparation
microbiology
storage temp.
2-8°C
Application
Preparation Note
Footnote
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.
Legal Information
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Salmonella, with 2,300+ serotypes, causes half of food-borne illnesses, often from dairy, poultry, and eggs.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service