Skip to Content
Merck
All Photos(1)

Key Documents

95273

Millipore

VRB MUG Agar

suitable for microbiology

Synonym(s):

Violet Red Bile Lactose MUG Agar

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106200
NACRES:
NA.74

Pricing and availability is not currently available.

shelf life

limited shelf life, expiry date on the label

Quality Level

composition

agar, 13 g/L
bile salt mixture, 1.5 g/L
crystal violet, 0.002 g/L
lactose, 10 g/L
meat peptone, 7 g/L
4-methylumbelliferyl-β-D-glucuronide, 0.1 g/L
neutral red, 0.03 g/L
sodium chloride, 5 g/L
yeast extract, 3 g/L

final pH

7.4±0.2 (25 °C)

application(s)

environmental
food and beverages
food and beverages
food and beverages

microbiology

suitability

selective and differential for Escherichia coli
selective and differential for Klebsiella spp.
selective and differential for Salmonella spp.
selective and differential for Shigella spp.
selective and differential for Yersinia spp.
selective and differential for coliforms
selective and differential for enterobacteriaceae

Application

Selective medium for the detection and enumeration of coliform bacteria, in particular E. coli. Gram-positive accompanying flora are extensively inhibited by crystalviolet and bile salts. A color-change to red indicates lactose-positive colonies, within which E. coli can be demonstrated by fluorescence in the UV.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

An article concerning selective growth media for differentiation and detection of Escherichia coli and other coliforms.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service