Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

AV48144

Sigma-Aldrich

Anti-TPI1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-MGC88108, Anti-TPI, Anti-Triosephosphate isomerase 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

27 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TPI1(7167)

General description

Triosephosphate isomerase 1 (TPI1) is an enzyme that catalyzes the isomerization of glyceraldehyde-3-phosphate and dihydroxy acetone phosphate during glycolysis and gluconeogenesis. Alterations in TPI1 gene have been linked to the deficiency of triosephosphate isomerase.
Rabbit Anti-TPI1 antibody recognizes bovine, pig, rabbit, human, and canine TPI1.

Immunogen

Synthetic peptide directed towards the N terminal region of human TPI1

Application

Rabbit Anti-TPI1 antibody has been used for western blot applications at a dilution of 1:1000.

Biochem/physiol Actions

TPI1 belongs to the triosephosphate isomerase family. Defects in TPI1 are the cause of triosephosphate isomerase deficiency (TPI deficiency) . TPI deficiency is an autosomal recessive disorder. It is the most severe clinical disorder of glycolysis. It is associated with neonatal jaundice, chronic hemolytic anemia, progressive neuromuscular dysfunction, cardiomyopathy and increased susceptibility to infection.

Sequence

Synthetic peptide located within the following region: MAPSRKFFVGGNWKMNGRKQSLGELIGTLNAAKVPADTEVVCAPPTAYID

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaoli Chen et al.
Reproduction (Cambridge, England), 147(3), 321-330 (2013-12-21)
Cryodamage is a major problem in semen cryopreservation, causing changes in the levels of proteins that influence the function and motility of spermatozoa. In this study, protein samples prepared from fresh and frozen-thawed boar spermatozoa were compared using the isobaric

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service