Skip to Content
Merck
All Photos(1)

Key Documents

HUMANPBMC-0002206

Sigma-Aldrich

Frozen Human Asthma PBMC

human peripheral blood (mononuclear cells)

Synonym(s):

Asthma PBMC, Immune Cells, Peripheral Blood Mononuclear Cells

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

UNSPSC Code:
41106514

Product Name

Frozen Human Asthma PBMC, 10 Million Cells/per vial

biological source

human peripheral blood (mononuclear cells)

Quality Level

form

frozen liquid

packaging

vial of 10 Million cells

manufacturer/tradename

BioIVT

technique(s)

cell culture | mammalian: suitable

color

cloudy

storage temp.

-140 to -196°C

General description

Lot specific orders are not able to be placed through the web. Contact your local sales rep for more details.

We are pleased to offer these products fromBioIVT.

Offering isolated Peripheral Blood Mononuclear Cells (PBMCs) from whole bloodprocured from IRB-consented donors with clinically defined Ulcerative Colitis.

Peripheral Blood Mononuclear Cells (PBMCs) are a subset of leukocytes (or WhiteBlood Cell, WBC), which are integral components of the body′s immune systemthat fight disease. PBMCs consist of a mixture of lymphocytes (T cells, Bcells, and NK cells) and monocytes and are isolated from peripheral whole bloodvia density gradient centrifugation and red blood cell lysis.

Standard and custom clinical data collection and confirmation ensures propercorresponding disease severity and treatment history.

Clinical data may include: Medication (current and historical), Diseaseseverity, Mayo Clinic Score, Montreal Score, Bristol Stool Type

PBMCs are provided with clinical data that has been collected following astrict and standardized clinical data process. All specimens are reviewed forIRB approval.

Packaging

10 Million Cells/per vial

Other Notes

Thaw before use.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

pictograms

Corrosion

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2

Storage Class

8B - Non-combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service