Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

R1401

Sigma-Aldrich

Reversin 205

≥98% (HPLC), film

Synonym(s):

[Boc-Glu(Obzl)]2-Lys-Ome

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C41H58N4O12
CAS Number:
Molecular Weight:
798.92
MDL number:
UNSPSC Code:
12161501
PubChem Substance ID:
NACRES:
NA.47

Quality Level

assay

≥98% (HPLC)

form

film

storage condition

desiccated

color

colorless

mp

101-102 °C

solubility

DMSO or ethanol: soluble (Insoluble in water)

storage temp.

−20°C

SMILES string

COC(=O)[C@H](CCCCNC(=O)[C@H](CCC(=O)OCc1ccccc1)NC(=O)OC(C)(C)C)NC(=O)[C@H](CCC(=O)OCc2ccccc2)NC(=O)OC(C)(C)C

InChI

1S/C41H58N4O12/c1-40(2,3)56-38(51)44-30(21-23-33(46)54-26-28-16-10-8-11-17-28)35(48)42-25-15-14-20-32(37(50)53-7)43-36(49)31(45-39(52)57-41(4,5)6)22-24-34(47)55-27-29-18-12-9-13-19-29/h8-13,16-19,30-32H,14-15,20-27H2,1-7H3,(H,42,48)(H,43,49)(H,44,51)(H,45,52)/t30-,31-,32-/m0/s1

InChI key

BRVAQYBFHAIYLQ-CPCREDONSA-N

Gene Information

human ... GRIN2B(2904)
mouse ... GRIN2B(14812)
rat ... GRIN2B(24410)

General description

Reversin 205 is a hydrophobic peptide.

Application

Reversin 205 has been used to determine the activity of P-glycoprotein in human retinal pigment epithelium.

Biochem/physiol Actions

Peptide chemosensitizer, inhibitor of P-glycoprotein.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Interaction of the P-glycoprotein multidrug transporter (MDR1) with high affinity peptide chemosensitizers in isolated membranes, reconstituted systems, and intact cells.
Sharom FJ
Biochemical Pharmacology, 58(4), 571-586 (1999)
P-glycoprotein expression in human retinal pigment epithelium
Brian G
Molecular Vision, 8 (2002)
Rohini Kondal et al.
Polymers, 13(17) (2021-09-11)
The impact of polymer-based slow-release urea formulations on soil microbial N dynamics in potatoes has been sparingly deciphered. The present study investigated the effect of a biodegradable nano-polymer urea formulation on soil enzymatic activities and microflora involved in the N

Articles

Cancer stem cell media, spheroid plates and cancer stem cell markers to culture and characterize CSC populations.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service