Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML1267

Sigma-Aldrich

Bryostatin 3

≥95% (HPLC)

Synonym(s):

[4S-[4R*(2E,4E),5R*,7E,9S*,11Z,13R*,15R*,17R*,19S*,21S*,25S*(S*),27S*,27aR*]]-2,4-Octadienoic acid,17-(acetyloxy)-4,5,6,9,10,11,12,13,14,15,16,17,18,19,20,21,22,23,25,26,27,27a-docosahydro-5,15,21-trihydroxy-25-(1-hydroxyethyl)-11-(2-methoxy-2-oxoethylidene)-6,6,16,16-tetramethyl-2,23-dioxo-5,27:9,13:15,19-triepoxy-2H-furo[2,3-e]oxacyclohexacosin-4-yl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C46H64O17
CAS Number:
Molecular Weight:
888.99
UNSPSC Code:
12352200
NACRES:
NA.77

biological source

Bugula neritina (sponge)

Quality Level

assay

≥95% (HPLC)

form

film

color

colorless to light yellow

shipped in

wet ice

storage temp.

−20°C

InChI

1S/C46H64O17/c1-9-10-11-12-13-14-37(50)61-42-33-23-40(53)60-41(33)35-24-34(26(2)47)59-39(52)21-29(49)20-31-22-36(57-27(3)48)44(6,7)45(54,62-31)25-32-18-28(19-38(51)56-8)17-30(58-32)15-16-43(4,5)46(42,55)63-35/h11-16,19,23,26,29-32,34-36,41-42,47,49,54-55H,9-10,17-18,20-22,24-25H2,1-8H3/b12-11+,14-13+,16-15-,28-19+/t26-,29-,30+,31-,32+,34-,35-,36+,41+,42+,45+,46-/m1/s1

InChI key

BSNHYLUEHJOXFN-UUIHWDITSA-N

Biochem/physiol Actions

Bryostatin 3 is a potent protein kinase C activator with a Ki of 2.75 nM, and an antagonist to phorbol ester tumor promoter TPA. Bryostatin 3 is isolated from the bryozoan Bugula neritina.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service