SRP0253
4EBP1 Active human
recombinant, expressed in E. coli, ≥70% (SDS-PAGE)
Synonym(s):
EIF4EBP1, Eukaryotic translation initiation factor 4E binding protein 1, PHAS-I, Phosphorylated heat- and acid-stable protein regulated by insulin 1, eIF4E-binding protein 1
About This Item
Recommended Products
biological source
human
recombinant
expressed in E. coli
assay
≥70% (SDS-PAGE)
form
aqueous solution
mol wt
13.6 kDa
packaging
pkg of 100 μg
storage condition
avoid repeated freeze/thaw cycles
concentration
>0.02 mg/mL
NCBI accession no.
UniProt accession no.
shipped in
dry ice
storage temp.
−70°C
Gene Information
human ... EIF4EBP1(1978)
General description
Human E1F4EBP1 (eukaryotic translation initiation factor 4E binding protein 1)(GenBank Accession No. NM_004095), full length with N-terminal His tag, MW = 13.6kDa, expressed in Escherichia coli.
Application
Biochem/physiol Actions
Physical form
Preparation Note
Storage Class
10 - Combustible liquids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service