MilliporeSigma
  • Promoter of lncRNA Gene PVT1 Is a Tumor-Suppressor DNA Boundary Element.
Promoter of lncRNA Gene PVT1 Is a Tumor-Suppressor DNA Boundary Element.
Cell (2018-05-08)
Seung Woo Cho, Jin Xu, Ruping Sun, Maxwell R Mumbach, Ava C Carter, Y Grace Chen, Kathryn E Yost, Jeewon Kim, Jing He, Stephanie A Nevins, Suet-Feung Chin, Carlos Caldas, S John Liu, Max A Horlbeck, Daniel A Lim, Jonathan S Weissman, Christina Curtis, Howard Y Chang
ABSTRACT
Noncoding mutations in cancer genomes are frequent but challenging to interpret. PVT1 encodes an oncogenic lncRNA, but recurrent translocations and deletions in human cancers suggest alternative mechanisms. Here, we show that the PVT1 promoter has a tumor-suppressor function that is independent of PVT1 lncRNA. CRISPR interference of PVT1 promoter enhances breast cancer cell competition and growth in vivo. The promoters of the PVT1 and the MYC oncogenes, located 55 kb apart on chromosome 8q24, compete for engagement with four intragenic enhancers in the PVT1 locus, thereby allowing the PVT1 promoter to regulate pause release of MYC transcription. PVT1 undergoes developmentally regulated monoallelic expression, and the PVT1 promoter inhibits MYC expression only from the same chromosome via promoter competition. Cancer genome sequencing identifies recurrent mutations encompassing the human PVT1 promoter, and genome editing verified that PVT1 promoter mutation promotes cancer cell growth. These results highlight regulatory sequences of lncRNA genes as potential disease-associated DNA elements.
Product Description
Roche
Cell Proliferation Kit I (MTT)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 g
Estimated to ship on March 23, 2025
$135.00
Sigma-Aldrich
Insulin from bovine pancreas, γ-irradiated, BioXtra, suitable for cell culture, potency: ≥20 units/mg (USP units), lyophilized powder
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 g
Estimated to ship on March 23, 2025
$135.00
Sigma-Aldrich
Cholera Toxin from Vibrio cholerae, ≥90% (SDS-PAGE), lyophilized powder
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 g
Estimated to ship on March 23, 2025
$135.00
Sigma-Aldrich
(+)-JQ1, ≥98% (HPLC)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 g
Estimated to ship on March 23, 2025
$135.00
Sigma-Aldrich
Insulin solution from bovine pancreas, 10 mg/mL insulin in 25  mM HEPES, pH 8.2, BioReagent, sterile-filtered, suitable for cell culture
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 g
Estimated to ship on March 23, 2025
$135.00
Sigma-Aldrich
Normal Mouse IgG, Normal Mouse IgG Polyclonal Antibody control validated for use in Immunoprecipitation & Western Blotting.
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 g
Estimated to ship on March 23, 2025
$135.00
Sigma-Aldrich
4-Thiouridine, ≥98%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 g
Estimated to ship on March 23, 2025
$135.00
Millipore
Accutase cell detachment solution, A cell detachment solution of proteolytic & collagenolytic enzymes. The reagent is useful for creating single cell suspensions from clumped cell cultures for accurate cell counting, detachment of cells from primary tissue.
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 g
Estimated to ship on March 23, 2025
$135.00